Skip to Content
Merck
All Photos(1)

Key Documents

230235

Sigma-Aldrich

Diphenylphosphinic chloride

98%

Synonym(s):

Diphenylchlorophosphine oxide, Diphenylphosphinic acid chloride, Diphenylphosphinoyl chloride, Diphenylphosphinyl chloride, Diphenylphosphorus oxychloride, Ph2POCl, Chlorodiphenylphosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2P(O)Cl
CAS Number:
Molecular Weight:
236.63
Beilstein:
975191
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

refractive index

n20/D 1.609 (lit.)

bp

222 °C/16 mmHg (lit.)

density

1.24 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClP(=O)(c1ccccc1)c2ccccc2

InChI

1S/C12H10ClOP/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

QPQGTZMAQRXCJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diphenylphosphinic chloride (Ph2POCl) is widely used as a precursor to synthesize various chiral phosphine bidentate ligands for asymmetric synthesis, in addition to several peptide coupling agents.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meifang Wang
Inorganic Chemistry, 33, 6, 1184-1187 (1994)
Difluorphos, an Electron?Poor Diphosphane: A Good Match Between Electronic and Steric Features.
Jeulin S, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 116(3), 324-329 (2004)
Atropisomeric amides as chiral ligands: Using (−)-sparteine-directed enantioselective silylation to control the conformation of a stereogenic axis.
Clayden J, et al.
The Journal of Organic Chemistry, 65(21), 7033-7040 (2000)
Indium?Mediated Asymmetric Allylation of Acylhydrazones Using a Chiral Urea Catalyst.
Tan K L and Jacobsen E N
Angewandte Chemie (International Edition in English), 46(8), 1315-1317 (2007)
Efficient syntheses of korupensamines A, B and michellamine B by asymmetric Suzuki-Miyaura coupling reactions.
Xu G, et al.
Journal of the American Chemical Society, 136(2), 570-573 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service