Skip to Content
Merck
All Photos(1)

Key Documents

01632

Sigma-Aldrich

Atto 520-Biotin

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

Biotin-Atto 520

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H53ClN6O8S
Molecular Weight:
777.37
MDL number:
UNSPSC Code:
12352108
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥90% (HPLC)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

solubility

DMF: soluble
ethanol: soluble
methanol: soluble

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 522-528 nm Amax

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[O-]Cl(=O)(=O)=O.CCNc1cc2OC3=CC(=[NH+]\CC)\C(C)=CC3=C(CCC(=O)NCCCCCNC(=O)CCCCC4SCC5NC(=O)NC45)c2cc1C

InChI

1S/C37H52N6O4S.ClHO4/c1-5-38-28-20-31-26(18-23(28)3)25(27-19-24(4)29(39-6-2)21-32(27)47-31)14-15-35(45)41-17-11-7-10-16-40-34(44)13-9-8-12-33-36-30(22-48-33)42-37(46)43-36;2-1(3,4)5/h18-21,30,33,36,38H,5-17,22H2,1-4H3,(H,40,44)(H,41,45)(H2,42,43,46);(H,2,3,4,5)/b39-29-;

InChI key

OCYRRKMPTKPPHX-QITHAKLPSA-N

Application

Atto 520 is a fluorescent label with high molecular absorption (110.000) and quantum yield (0.90) as well as sufficient stoke′s shift (excitation maximum 520 nm, emission maximum 542 nm). It does nearly not show cis-trans-isomerisation, which limits brightness and reproducibility for many other dyes. Due to an insignificant triplet formation rate it is well suited for single molecule detection applications. Can be utilized in applications such as ELISA or immuno-histochemistry, in situ hybridization, flow cytometry and others, to identify streptavidin, avidin, or extravidin-conjugates. Atto-520 fluorescent-labeled biotin can be loaded onto magnetic beads coated with streptavidin through specific ligand–receptor interactions.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manoj Kumbhakar et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 10(4), 629-633 (2009-01-30)
Complex dynamics of combined long-range fluorescence resonance energy transfer (FRET) and short-range electron transfer (ET) processes in a single double-stranded DNA (dsDNA) molecule (see figure) reveals that FRET remains almost unaltered in the presence of ET. Present systems also demonstrate
Vladimir Mekler et al.
The Journal of biological chemistry, 286(1), 270-279 (2010-10-19)
Promoter recognition by RNA polymerase is a key point in gene expression and a target of regulation. Bacterial RNA polymerase binds promoters in the form of the holoenzyme, with the σ specificity subunit being primarily responsible for promoter recognition. Free
Hee-Jin Jeong et al.
Biosensors & bioelectronics, 40(1), 17-23 (2012-07-17)
Protein phosphorylation is a key event in intracellular signal transduction, and fluorescent biosensor for the specific phosphorylation event in a target protein is considered highly useful as a tool of cellular biology and drug screening. Vimentin, the most abundant intermediate
Christopher A Teske et al.
The journal of physical chemistry. B, 109(28), 13811-13817 (2006-07-21)
Confocal laser scanning microscopy (CLSM) is being increasingly used for observing protein uptake in porous chromatography resins. Recent CLSM studies have revealed the possible existence of a nondiffusive protein transport mechanism. Observing protein uptake with CLSM requires labeling the protein
Siddharth Nanguneri et al.
PloS one, 7(5), e38098-e38098 (2012-06-05)
Three-dimensional fluorescence imaging of thick tissue samples with near-molecular resolution remains a fundamental challenge in the life sciences. To tackle this, we developed tomoSTORM, an approach combining single-molecule localization-based super-resolution microscopy with array tomography of structurally intact brain tissue. Consecutive

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service