Skip to Content
Merck
All Photos(1)

Key Documents

S9663

Sigma-Aldrich

Sodium arsenate dibasic heptahydrate

ACS reagent, ≥98%

Synonym(s):

Disodium hydrogen arsenate heptahydrate, di-Sodium hydrogen arsenate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HAsO4 · 7H2O
CAS Number:
Molecular Weight:
312.01
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥98%
98.0-102.0%
grade:
ACS reagent
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥98%
98.0-102.0%

form

powder

impurities

≤0.005% Insoluble matter

pH

8.5-9 (25 °C, 50 g/L)

anion traces

arsenite (As2O3): ≤0.01%
chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.005%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O

InChI

1S/AsH3O4.2Na.7H2O/c2-1(3,4)5;;;;;;;;;/h(H3,2,3,4,5);;;7*1H2/q;2*+1;;;;;;;/p-2

InChI key

KOLXPEJIBITWIQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium arsenate dibasic heptahydrate can be used as an internal standard for the quantification of arsenic species using various spectroscopic methods.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shugo Suzuki et al.
Toxicology, 299(2-3), 155-159 (2012-06-06)
Inorganic arsenic is a known human carcinogen, inducing tumors of the skin, urinary bladder and lung. It is metabolized to organic methylated arsenicals. 2,3-Dimercaptopropane-1-sulfonic acid (DMPS), a chelating agent, is capable of reducing pentavalent arsenicals to the trivalent state and
M Sri Lakshmi Sunita et al.
Ecotoxicology (London, England), 21(1), 202-212 (2011-09-01)
In the present study, 44 arsenic-resistant bacteria were isolated through serial dilutions on agar plate with concentrations ≥0.05 mM of sodium arsenite and ≥10 mM of sodium arsenate from Mandovi and Zuari--estuarine water systems. The ars genotype characterization in 36
Tatyana S Pinyayev et al.
Chemical research in toxicology, 24(4), 475-477 (2011-03-11)
The conventional scheme for arsenic methylation accounts for methylated oxyarsenical production but not for thioarsenical formation. Here, we report that in vitro anaerobic microbiota of mouse cecum converts arsenate into oxy- and thio- arsenicals. Besides methylarsonic acid (MMA(V)), arsenate was
Fariha Qureshi et al.
Journal of Ayub Medical College, Abbottabad : JAMC, 21(4), 63-69 (2010-11-12)
Arsenic is a teratogenic agent present in the environment as oxides and arsenate and humans are exposed to it through contaminated drinking water, food, soil and air. This investigation was undertaken to evaluate protective role of Vitamin C and E
William Brattin et al.
Journal of toxicology and environmental health. Part A, 76(7), 449-457 (2013-04-25)
This study describes a method for measuring the relative oral bioavailability (RBA) of arsenic (As) in soil and other soil-like media using young swine as the animal model. Groups of animals are exposed to site soil or sodium arsenate orally

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service