Skip to Content
Merck
All Photos(1)

Key Documents

83381

Sigma-Aldrich

Mercury(II) nitrate monohydrate

puriss. p.a., ACS reagent, ≥98.5% (T)

Synonym(s):

Mercuric nitrate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(NO3)2 · H2O
CAS Number:
Molecular Weight:
342.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥98.5% (T)
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥98.5% (T)

form

powder or crystals

ign. residue (after reduction)

≤0.01%

mp

79 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
Hg+: ≤2000 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Hg.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

KVICROHOONHSRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An ion-imprinted polymer for the selective extraction of mercury (II) ions in aqueous media.
Batlokwa BS, et al.
Water SA , 38(2), 255-260 (2012)
Factors affecting Hg (II) adsorption on hybrid nanostructured silicas: influence of the synthesis conditions.
Perez-Quintanilla D and Sierra I.
Journal of Porous Materials, 21(1), 71-80 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service