Skip to Content
Merck
All Photos(2)

Key Documents

E12508

Sigma-Aldrich

Ethylbenzene

ReagentPlus®, 99%

Synonym(s):

NSC 406903, Phenylethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C2H5
CAS Number:
Molecular Weight:
106.17
Beilstein:
1901871
EC Number:
MDL number:
UNSPSC Code:
12352002
eCl@ss:
39011201
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.7 (vs air)

Quality Level

vapor pressure

10 mmHg ( 20 °C)
19 mmHg ( 37.7 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

810 °F

expl. lim.

6.7 %

refractive index

n20/D 1.495 (lit.)

bp

136 °C (lit.)

mp

−95 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylbenzeneis a clear liquid with a noticeable aromatic fragrance. It is flammable and easily blends with alcohol and ether, although it has a low solubility in water. It is commonly used as a solvent and vital in the production of styrene and synthetic polymers, which are employed in the manufacturing of plastics and rubber items.

Application

Ethylbenzenecan be used as a solvent to synthesis Pseudo-Peptides or N,N-disubstituted thiomalonamides with retro thioamide bond.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT RE 2

Target Organs

hearing organs

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Pseudo-Peptide Building Blocks with retro-Thioamide Bond Mediated via Thiocarbamoyl Meldrum's Acid
Janikowska K, et al.
Helvetica Chimica Acta, 95(3), 461-468 (2012)
Enza Palma et al.
New biotechnology, 53, 41-48 (2019-07-01)
Microbial electrochemical technologies (MET) are increasingly being considered for in situ remediation of contaminated groundwater. However, their application potential for the simultaneous treatment of complex mixtures of organic and inorganic contaminants, has been only marginally explored. Here we have analyzed
Jose Ruiz-Jimenez et al.
Journal of chromatography. A, 1616, 460825-460825 (2020-01-12)
Several calibration approaches were evaluated for the quantitation of volatile organic compounds in air using miniaturized exhaustive and non-exhaustive sampling techniques, such as in-tube extraction (ITEX) and solid phase microextraction (SPME) Arrow. Eleven compounds, 2-ethyl-hexanol, hexanal, nonanal, toluene, ethyl-benzene, methyl
Lili Zhang et al.
Chemosphere, 90(4), 1340-1347 (2012-09-11)
A new strain Mycobacterium cosmeticum byf-4 able to simultaneously degrade benzene, toluene, ethylbenzene, and o-xylene (BTE(o-)X) compounds has been isolated and identified previously in our laboratory. We further report here the extent of degradation of every BTE(o-)X component, and unravel
Carla G A da Silva et al.
Journal of chromatography. A, 1260, 81-87 (2012-09-12)
Endcapped stationary phases were prepared after thermal immobilization of poly(methyloctadecylsiloxane) (PMODS) onto zirconized and titanized silica supports. These new stationary phases have lower densities of residual hydroxyl groups, according to infrared spectroscopy and 29Si CP-MAS NMR and as shown by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service