C87657
Cumene
98%
Synonym(s):
(1-Methylethyl)benzene, 2-Phenylpropane, Isopropylbenzene, NSC 8776
About This Item
Recommended Products
vapor density
4.1 (vs air)
vapor pressure
8 mmHg ( 20 °C)
9.7 mmHg ( 37.7 °C)
Assay
98%
form
liquid
autoignition temp.
797 °F
expl. lim.
6.5 %
refractive index
n20/D 1.491 (lit.)
bp
152-154 °C (lit.)
mp
−96 °C (lit.)
density
0.864 g/mL at 25 °C (lit.)
SMILES string
CC(C)c1ccccc1
InChI
1S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8H,1-2H3
InChI key
RWGFKTVRMDUZSP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Diisopropylbenzenes via disproportionation reaction by zeolite catalysts.
- Cumene hydroperoxide, a key precursor for many organic fine chemicals.
- 4-Nitro cumene using nitric acid/ MoO3/SiO2 as a catalyst.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 3 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
87.8 °F - closed cup
Flash Point(C)
31.0 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service