Skip to Content
Merck
All Photos(3)

Key Documents

D65100

Sigma-Aldrich

Dichloromethane

contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%

Synonym(s):

methylene chloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04
grade:
ACS reagent
Assay:
≥99.5%
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

grade

ACS reagent

Quality Level

100
200

Agency

suitable for EPA 1613

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

1223 °F

contains

40-150 ppm amylene as stabilizer

expl. lim.

22 %

impurities

Free halogens, passes test
≤0.0003 meq/g Titr. acid
≤0.02% water

evapn. residue

≤0.002%

color

APHA: ≤10
clear

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane (DCM) is a chlorinated organic solvent with a wide range of industrial applications and organic syntheses. It has a low boiling point, high density and is immiscible in water. DCM is susceptible to degradation with time, which can be suppressed by adding amylene as a stabilizer.

Application

Dichloromethane
  • Organic synthesis
a) Dakin, Dess-Martin and Ley Oxidation
b) Enyne metathesis, widely used catalyst is the Grubbs′s second-generation ruthenium complex
c) Jacobsen-Katsuki epoxidation
d) Boc-deprotection of immobilized Boc-NH2-poly(ethylene glycol)-plasma layers
e) Can react with phosphines or amines in the formation of the corresponding alkylated products
  • Adhesive and paint formulation
  • Extraction-concentration analysis
  • Flash chromatograph
  • Electronic industry in printed circuits board
  • Production of cellulose triacetate

Features and Benefits

ACS solvents meet or exceed the high standards of the American Chemical Society (ACS), with test specifications that are specialized to every compound. According to the American Chemical Society, ACS reagent grade implies that it is a substance of sufficient purity to be used in most chemical analyses or reactions

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Instrument and technique for the in vitro screening of platelet activation from whole blood samples.
Martin Y, et al.
The Review of Scientific Instruments, 78(5), 054302-054302 (2007)
Biodegradation of dichloromethane in an estuarine environment.
Krausova VI, et al.
Hydrobiologia, 559(1), 77-83 (2006)
Dichloromethane Detection Based on Near-Infrared Absorptive Sensing.
Han JH and Yoon SM
IEEE Journal of Selected Topics in Quantum Electronics, 18(5), 1547-1552 (2012)
Vitamin B12-mediated hydrodechlorination of dichloromethane by bimetallic Cu/Al particles.
Huang CC, et al.
Chemical Engineering Journal, 273, 413-420 (2015)
Dichloromethane photodegradation using titanium catalysts.
Tanguay JF, et al.
Tanguay JF, et al.
J. Catal., 117(2), 335-347 (1989)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service