Skip to Content
Merck
All Photos(2)

Key Documents

D201863

Sigma-Aldrich

1,4-Dioxane

ReagentPlus®, ≥99%, contains ≤25 ppm BHT as stabilizer

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
technique(s):
thin layer chromatography (TLC): suitable
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

grade

reagent

Quality Level

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

356 °F

contains

≤25 ppm BHT as stabilizer

expl. lim.

22 %

technique(s)

thin layer chromatography (TLC): suitable

refractive index

n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Dioxane can be used as a solvent:
  • To synthesize 1,3-diamines and amino alcohols via 2-azaallyl anion benzylic ring-opening of aziridines.
  • In the RAFT polymerization of acryclic acid.
  • In the synthesis of cross-linked triblock copolymer hydrogels.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-toughness polycation cross-linked triblock copolymer hydrogels
Chen Y, et al.
Macromolecules, 50, 3637-3646 (2017)
Photoactive rose bengal-based latex via RAFT emulsion polymerization-induced self-assembly
Boussiron C, et al.
Open Journal of Polymer Chemistry, 12, 134-147 (2021)
Yimu Zhao et al.
Advanced healthcare materials, 8(5), e1801187-e1801187 (2019-02-10)
Due to escalating drug developmental costs and limitations of cardiotoxicity screening, there is an urgent need to develop robust in vitro 3D tissue culture platforms that can both facilitate the culture of human cardiac tissues and provide noninvasive functional readouts
Christof Aellig et al.
ChemSusChem, 5(9), 1737-1742 (2012-07-05)
The dehydration of D-fructose to 5-hydroxymethylfurfural was studied under single-phase conditions in the low boiling solvent 1,4-dioxane at moderate temperatures in the presence of the solid acid-catalyst Amberlyst-15. The reaction was first examined and optimized under batch conditions, where it
Alkene trifluoromethylation coupled with C-C bond formation: construction of trifluoromethylated carbocycles and heterocycles.
Hiromichi Egami et al.
Angewandte Chemie (International ed. in English), 52(14), 4000-4003 (2013-02-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service