Skip to Content
Merck
All Photos(3)

Key Documents

360481

Sigma-Aldrich

1,4-Dioxane

ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥99.0%
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

grade

ACS reagent

Quality Level

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

356 °F

contains

≤25 ppm BHT as stabilizer

expl. lim.

22 %

impurities

≤0.0016 meq/g Titr. acid
≤0.005% Peroxide (as H2O2)
≤0.01% Carbonyl (as HCHO)
≤0.05% water

evapn. residue

≤0.005%

color

APHA: ≤20

refractive index

n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

transition temp

freezing point ≥11.0

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dioxane is a heterocyclic organic compound that is commonly used as a solvent in several chemical reactions. It is also used as a stabilizer in chlorinated solvents.

Application

1,4-Dioxane can be used as:      
  • A solvent in the N-methylation of amines by reductive functionalization of CO2 in the presence of NaBH4 as the reducing reagent.      
  • An additive to synthesize 3-hydroxylisoindolin-1-ones via hydroxylhydrative aza-cyclization of 2-alkynylbenzamide.

A mixture of 1,4-dioxane/ethanol/formic acid solvent system can be used in the depolymerization of lignin to synthesize value-added phenolic monomers.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel J Williamson et al.
Nature protocols, 9(2), 253-262 (2014-01-11)
Technologies that allow the efficient chemical modification of proteins under mild conditions are widely sought after. Sortase-mediated peptide ligation provides a strategy for modifying the N or C terminus of proteins. This protocol describes the use of depsipeptide substrates (containing
Oxidation and biodegradability enhancement of 1,4-dioxane using hydrogen peroxide and ozone.
C D Adams et al.
Environmental science & technology, 28(11), 1812-1818 (1994-10-01)
Self-curable benzoxazine functional polybutadienes synthesized by click chemistry.
Kukut M, et al.
Designed Monomers and Polymers, 12(2), 167-176 (2009)
Kellan T Passow et al.
Current protocols in nucleic acid chemistry, 80(1), e101-e101 (2020-01-08)
4-Cyanoindole-2'-deoxyribonucleoside (4CIN) is a fluorescent isomorphic nucleoside analogue with superior spectroscopic properties in terms of Stokes shift and quantum yield in comparison to the widely utilized isomorphic nucleoside analogue, 2-aminopurine-2'-deoxyribonucleoside (2APN). Notably, when inserted into single- or double-stranded DNA, 4CIN
Christof Aellig et al.
ChemSusChem, 5(9), 1737-1742 (2012-07-05)
The dehydration of D-fructose to 5-hydroxymethylfurfural was studied under single-phase conditions in the low boiling solvent 1,4-dioxane at moderate temperatures in the presence of the solid acid-catalyst Amberlyst-15. The reaction was first examined and optimized under batch conditions, where it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service