Skip to Content
Merck
All Photos(1)

Key Documents

96459

Sigma-Aldrich

Zinc acetate dihydrate

puriss. p.a., ACS reagent, ≥99.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
219.51
Beilstein:
3732513
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0% (KT)

form

crystals

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤0.002% total nitrogen (N)

pH

6-7

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg

SMILES string

O.O.CC(=O)O[Zn]OC(C)=O

InChI

1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2

InChI key

BEAZKUGSCHFXIQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc acetate dihydrate may be employed as a chelating agent in the synthesis of nanocrystalline ZnO powders. It may be employed in the synthesis of high-purity single crystal ZnO nanowires, via thermal decomposition at 300°C in air.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of zinc acetate dihydrate, Zn(CH3COO)2.2H2O.
Van Niekerk JN, et al.
Acta Crystallographica, 6(8), 720-720 (1953)
Odair Bim-Júnior et al.
ACS biomaterials science & engineering, 6(8), 4539-4550 (2021-01-19)
Plant-derived compounds incite applications virtually on every biomedical field due to the expedient antioxidant, anti-inflammatory and antimicrobial properties in conjunction with a natural character. Here, quercetin (QCT), a flavonoid with therapeutic potentials relevant to the oral environment, was encapsulated within
Synthesis of ZnO nanowires by thermal decomposition of zinc acetate dihydrate.
Lin CC and Li YY.
Materials Chemistry and Physics, 113(1), 102-106 (2009)
Synthesis and optical properties of nanocrystalline ZnO powders by a simple method using zinc acetate dihydrate and poly (vinyl pyrrolidone).
Maensiri S, et al.
Journal of Crystal Growth, 289(1), 102-106 (2006)
Robert Krysiak et al.
The Yale journal of biology and medicine, 85(2), 249-254 (2012-06-28)
Wilson's disease is a rare genetic disorder of copper metabolism. The difference in copper tissue accumulation is responsible for the various clinical manifestations of this disorder. If left untreated, Wilson's disease progresses to hepatic failure, severe neurological disability, and even

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service