Skip to Content
Merck
All Photos(1)

Key Documents

411000

Sigma-Aldrich

Ethanolamine

purified by redistillation, ≥99.5%

Synonym(s):

2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
Beilstein:
505944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.1 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

1436 °F

purified by

glass distillation
redistillation

expl. lim.

17 %

refractive index

n20/D 1.454 (lit.)

bp

170 °C (lit.)
69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanolamine is an alkanolamine. Ethanolamine is an essential growth-promoting supplement for serum-free medium for culturing mouse plasmacytomas and hybridoma cell lines of MPC11-BL and SP2/0-BL origins. It is reported to efficiently inhibit the coupling reaction of aryl halides.

Application

Ethanolamine was used for the synthesis of N-oleoylaminoethanol standard. It was used for the preparation and derivatization of poly(acrylic acid) (PAA) films. It may be used in scintillating method for evaluating counts for carbon-14.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid scintillation counting of carbon-14. Use of ethanolamine-ethylene glycol monomethyl ether-toluene.
Jeffay H and Alvarez J.
Analytical Chemistry, 33(4), 612-615 (1961)
H Murakami et al.
Proceedings of the National Academy of Sciences of the United States of America, 79(4), 1158-1162 (1982-02-01)
A serum-free medium supplemented with a few growth factors was devised to grow lymphocyte hybridomas. The medium was developed with the hybridoma line MPC11-BL, a fusion product between a mouse plasmacytoma cell line (MPC11TG70na3) and mouse (BALB/c) spleen cells. In
Aqueous solvation and functionalization of weak-acid polyelectrolyte thin films.
Peez RF, et al.
Langmuir, 14(15), 4232-4237 (1998)
Surface reactions of brominated arenes as a model for the formation of chlorinated dibenzodioxins and-furans in incineration: inhibition by ethanolamine.
Lippert T, et al.
Environmental Science & Technology, 25(8), 1485-1489 (1991)
Fong-Fu Hsu et al.
Journal of the American Society for Mass Spectrometry, 13(5), 558-570 (2002-05-22)
Negative-ion electrospray ionization tandem quadrupole mass spectrometry provides a useful method for the structural characterization of ceramides. Fragment ions referring to the identities of the fatty acid substituent and of the long chain base of the molecules are readily available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service