Skip to Content
Merck
All Photos(1)

Key Documents

85260

Sigma-Aldrich

Silver(I) oxide

purum p.a., ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2O
CAS Number:
Molecular Weight:
231.74
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

≥99.0% (AT)

form

powder

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Ag]O[Ag]

InChI

1S/2Ag.O

InChI key

KHJDQHIZCZTCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver(I) oxide reacts with CO2 present in the atmosphere to afford silver carbonate. It is widely used in the preparation of ceramic pigments. It participates in the Arndt-Eistert synthesis.

Application

Silver(I) oxide may be used in the synthesis of following:
  • (RS)-malic acid
  • 1,2-(o)-benzoquinone
  • polysubstituted cyclic ethers

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abderrahim Bouzide et al.
Organic letters, 4(14), 2329-2332 (2002-07-06)
[reaction: see text] The reaction of symmetrical diols and oligo(ethylene glycol)s with a stoichiometric amount of p-toluenesulfonyl chloride in the presence of silver(I) oxide and a catalytic amount of potassium iodide led selectively to the monotosylate derivatives in high yields.
Eagleson M.
Concise Encyclopedia Chemistry, 90-90 (1994)
Chiara Uboldi et al.
Mutation research, 745(1-2), 11-20 (2011-11-19)
Although amorphous silica nanoparticles (aSiO(2)NPs) are believed to be non-toxic and are currently used in several industrial and biomedical applications including cosmetics, food additives and drug delivery systems, there is still no conclusive information on their cytotoxic, genotoxic and carcinogenic
Y Yonekura et al.
The Journal of bone and joint surgery. British volume, 93(5), 644-649 (2011-04-23)
A silver-containing hydroxyapatite (Ag-HA) coating has been developed using thermal spraying technology. We evaluated the osteoconductivity of this coating on titanium (Ti) implants in rat tibiae in relation to bacterial infection in joint replacement. At 12 weeks, the mean affinity
Arixin Bo et al.
Journal of hazardous materials, 246-247, 199-205 (2013-01-15)
Emergency treatment of radioactive material leakage and safety disposal of nuclear waste is a constant concern all along with the development of radioactive materials applications. To provide a solution, titanate with large surface area (143 m(2)g(-1)) and a lamina morphology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service