Skip to Content
Merck
All Photos(1)

Key Documents

79613

Sigma-Aldrich

meta-Phosphoric acid

puriss. p.a., ACS reagent, ≥33.5% (T)

Synonym(s):

Metaphosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HPO3)n
CAS Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

form

crystalline

contains

57-63% NaPO3 as stabilizer

concentration

≥33.5% (T)

technique(s)

HPLC: suitable

solubility

water: miscible

density

2 g/cm3

anion traces

chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OP(=O)=O

InChI

1S/HO3P/c1-4(2)3/h(H,1,2,3)

InChI key

UEZVMMHDMIWARA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

meta-Phosphoric acid is a monobasic acid.

Application

meta-Phosphoric acid may be employed as an extraction reagent during isocratic reversed-phase ion-pairing high performance liquid chromatographic analysis of ascorbic acid and dehydroascorbic acid in plant.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Kall et al.
Journal of chromatography. B, Biomedical sciences and applications, 730(1), 101-111 (1999-08-07)
The total vitamin C amount in different food and plasma samples was determined by a dual detection system, after HPLC separation, with direct detection of ascorbic acid and indirect fluorimetric detection of dehydroascorbic acid after a post-column O-phenyldiamine derivatisation. The

Articles

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service