Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
meta-Phosphoric acid
Synonym(s):
Metaphosphoric acid
Linear Formula:
(HPO
3
)
n
CAS Number:
37267-86-0
EC Number:
253-433-4
Products
Product Category
inorganic acids (4)
Brand
Sigma-Aldrich (4)
Physical Form
Product Line
Quality Segment
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Purum (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
239275
meta-Phosphoric acid,
ACS reagent, chips, 33.5-36.5%
Expand
View Pricing
79615
meta-Phosphoric acid,
purum, ~65% HPO
3
basis, pieces
Expand
View Pricing
79613
meta-Phosphoric acid,
puriss. p.a., ACS reagent, ≥33.5% (T)
Expand
View Pricing
M6288
meta-Phosphoric acid,
BioXtra, ≥33.5%
Expand
View Pricing