Skip to Content
Merck
All Photos(1)

Key Documents

71504

Sigma-Aldrich

Sodium phosphate monobasic monohydrate

puriss. p.a., ACS reagent, ≥99.0% (T)

Synonym(s):

Monosodium phosphate, Sodium dihydrogen phosphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0% (T)

form

crystals

impurities

≤0.001% total nitrogen (N)
≤0.01% insoluble matter

pH

4.1-4.5 (25 °C, 50 mg/mL in H2O)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

[Na+].[H]O[H].OP(O)([O-])=O

InChI

1S/Na.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+1;;/p-1

InChI key

BBMHARZCALWXSL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate monobasic monohydrate is a sodium salt.

Application

Sodium phosphate monobasic monohydrate may be used in the following studies:
  • As a mobile phase in HPLC studies.
  • For the dissolution of BSA (Bovine serum albumin).
  • Preparation of buffer solutions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Lopes-de-Araújo et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 123, 515-523 (2018-08-08)
Topotecan is a relatively large, planar, asymmetric and polar molecule with a lactone moiety. In neutral or basic aqueous solutions, this ring opens forming the carboxylate form of Topotecan that is biologically inactive and uncapable of passively cross membranes. Nevertheless
P Yi et al.
The Journal of biological chemistry, 275(38), 29318-29323 (2000-07-08)
S-Adenosylmethionine and S-adenosylhomocysteine (SAH), as the substrate and product of essential cellular methyltransferase reactions, are important metabolic indicators of cellular methylation status. Chronic elevation of SAH, secondary to the homocysteine-mediated reversal of the SAH hydrolase reaction, reduces methylation of DNA
N Tsapis et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(19), 12001-12005 (2002-08-30)
We have combined the drug release and delivery potential of nanoparticle (NP) systems with the ease of flow, processing, and aerosolization potential of large porous particle (LPP) systems by spray drying solutions of polymeric and nonpolymeric NPs into extremely thin-walled
Inkyu Park et al.
Biosensors & bioelectronics, 22(9-10), 2065-2070 (2006-10-24)
A microneedle sensor platform with integrated silicon nanowire tip was developed for intracellular biochemical detection. Because of the virtue of miniaturized size and high sensitivity, this sensor has a great potential for studying individual cell or localized bioenvironment by revealing
Islem Gandouzi et al.
Materials (Basel, Switzerland), 12(7) (2019-04-14)
Pyoverdine is a fluorescent siderophore produced by Pseudomonas aeruginosa that can be considered as a detectable marker in nosocomial infections. The presence of pyoverdine in water can be directly linked to the presence of the P. aeruginosa, thus being a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service