Skip to Content
Merck
All Photos(2)

Key Documents

431478

Sigma-Aldrich

Sodium phosphate dibasic heptahydrate

≥99.99% trace metals basis

Synonym(s):

Disodium hydrogen phosphate heptahydrate, Disodium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 7H2O
CAS Number:
Molecular Weight:
268.07
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

vapor density

4.9 (vs air)

Assay

≥99.99% trace metals basis

form

solid

impurities

≤0.005% insolubles
<100 ppm total metallic impurities

pH

8.7-9.3

density

1.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals: ≤0.001%

functional group

phosphate

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.7H2O/c;;1-5(2,3)4;;;;;;;/h;;(H3,1,2,3,4);7*1H2/q2*+1;;;;;;;;/p-2

InChI key

PYLIXCKOHOHGKQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate dibasic heptahydrate has been used to prepare sodium phosphate buffer and phosphate-buffered saline (PBS) buffer.[44}

Features and Benefits

Meets A.C.S. reagent specifications.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial Competition Assay Based on Extracellular D-amino Acid Production.
Alvarez L
Bio-protocol, 8(7), e2787-e2787 (2018)
Walter Grünberg et al.
The British journal of nutrition, 110(6), 1012-1023 (2013-04-03)
Hypophosphataemia is frequently encountered in dairy cows during early lactation. Although supplementation of P is generally recommended, controversy exists over the suitability of oral P supplementation in animals with decreased or absent rumen motility. Since the effects of transruminal P
M A Marzouk et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 114, 658-667 (2013-07-03)
The effects of gamma irradiation on spectral properties of Cr2O3-doped phosphate glasses of three varieties, namely sodium metaphosphate, lead metaphosphate and zinc metaphosphate have been investigated. Optical spectra of the undoped samples reveal strong UV absorption bands which are attributed
Matthew H Crouthamel et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(11), 2625-2632 (2013-08-24)
Elevated serum phosphate has emerged as a major risk factor for vascular calcification. The sodium-dependent phosphate cotransporter, PiT-1, was previously shown to be required for phosphate-induced osteogenic differentiation and calcification of cultured human vascular smooth muscle cells (VSMCs), but its
H Meredith et al.
International journal of food microbiology, 166(2), 309-315 (2013-08-27)
Campylobacteriosis is the most common foodborne bacterial infection in developed countries and many cases are associated with poultry. This study investigated the immediate and storage effect of dipping inoculated poultry skin samples in trisodium phosphate (TSP, 10 & 14%, w/v)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service