Skip to Content
Merck
All Photos(1)

Key Documents

60370

Sigma-Aldrich

Potassium hydroxide

puriss. p.a., ≥86% (T), pellets

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥86% (T)
form:
pellets
solubility:
alcohol: soluble(lit.)
water: soluble(lit.)
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 719 °C)

Quality Level

grade

puriss. p.a.

Assay

≥86% (T)

form

pellets

impurities

≤0.0005% total nitrogen (N)
≤0.6% potassium carbonate (K2CO3)

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

solubility

alcohol: soluble(lit.)
water: soluble(lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
phosphate (PO43-): ≤5 mg/kg
silicate (as SiO2): ≤5 mg/kg
sulfate (SO42-): ≤40 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Hg: ≤0.1 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤5000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide (KOH, Caustic potash) is a base. It is a water absorbing, colorless solid having rhombic crystals. It has been synthesized by reacting potassium carbonate with calcium hydroxide. It is widely employed for the manufacture of dyes and pigments.[1] Pulverized KOH may be used for the preparation of chalcones.[2]

Application

Potassium hydroxide (KOH) may be employed for the following studies:
  • As an etching agent for the laser modified silica glass. It selectively facilitates etching on exposure to femtosecond laser.[3]
  • As a catalyst for the transformation of palm oil along with methanol to methyl esters, via transesterification reaction.[4]
  • Preparation of potassium hydroxide catalyst supported on palm shell activated carbon, which is used for the transesterification of palm oil.[5]
  • To improve the catalytic efficacy of X, Y and ZSM (Zeolite Socony Mobil)-5 zeolites during the synthesis of methyl methacrylate.[6]

Other Notes

Reagent for the decomposition of silicates by fusion[7][8]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Tien Duc Pham et al.
Polymers, 12(1) (2020-01-08)
The present study aims to investigate adsorptive removal of molecular ciprofloxacin using protein-modified nanosilica (ProMNS). Protein was successfully extracted from Moringa seeds while nanosilica was synthesized from rice husk. Fourier-transform infrared (FTIR), ultraviolet visible (UV-Vis) and high-performance liquid chromatography (HPLC)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Rokas Drevinskas et al.
Optics express, 23(2), 1428-1437 (2015-04-04)
Surface texturing is demonstrated by the combination of wet etching and ultrafast laser nanostructuring of silica glass. Using potassium hydroxide (KOH) at room temperature as an etchant of laser modified glass, we show the polarization dependent linear increase in retardance
Aldol condensation in gaseous phase by zeolite catalysts.
Wierzchowski PT and Zatorski LW.
Catalysis Letters, 9(5-6), 411-414 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service