Skip to Content
Merck
All Photos(1)

Key Documents

60239

Sigma-Aldrich

Potassium fluoride

puriss. p.a., ≥99.0% (F)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein:
3902818
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.0% (F)
form:
powder or crystals

grade

puriss. p.a.

Quality Level

Assay

≥99.0% (F)

form

powder or crystals

impurities

≤0.01% free acid (as HF)
≤0.01% free alkali (as KOH)
≤0.1% potassium fluoro silicate (K2SiF6)

mp

858 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤5000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium fluoride (KF) on activated alumina is a highly reactive catalyst (basic in nature) for various organic reactions. High reactivity is due to the formation of potassium hexafluoroaluminiate, potassium hydroxide and potassium aluminate on the interaction of its fluoride ion with alumina. Celite coated with potassium fluoride participates in various hydrogen-bond-mediated reactions. Potassium fluoride on basic alumina in acetonitrile has been employed for the ultrasound-assisted deprotection of tert-butyldimethylsilyl ethers of phenols. Neutralization reaction of hydrofluoric acid with KOH or postassium carbonate affords KF.

Application

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.

Other Notes

Reagent for the decomposition of silicates by fusion

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Characterization of the actual catalytic agent in potassium fluoride on activated alumina systems.
Weinstock LM, et al.
Tetrahedron Letters, 27(33), 3845-3848 (1986)
Natural phosphate doped with potassium fluoride and modified with sodium nitrate: Efficient catalysts for the Knoevenagel condensation.
Sebti S, et al.
Tetrahedron Letters, 43(10), 1813-1815 (2002)
Selective desilylation of tert-butyldimethylsilyl ethers of phenols using potassium fluoride-alumina and ultrasound.
Schmittling EA and Sawyer JS.
Tetrahedron Letters, 32(49), 7207-7210 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service