Skip to Content
Merck
All Photos(1)

Key Documents

1.04994

Supelco

Potassium fluoride

for analysis EMSURE® ACS

Synonym(s):

Potassium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FK
CAS Number:
Molecular Weight:
58.10
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-151-5
NACRES:
NA.21
Assay:
≥99.0% (precipitation titration)
grade:
ACS reagent
form:
solid
solubility:
923 g/L

grade

ACS reagent

Quality Level

vapor pressure

1.3 hPa ( 885 °C)

product line

EMSURE®

Assay

≥99.0% (precipitation titration)

form

solid

potency

245 mg/kg LD50, oral (Rat)

impurities

≤0.05% Free acid (as HF)

pH

8-9 (20 °C, 50 g/L in H2O)

bp

1505 °C/1013 hPa

mp

846 °C

solubility

923 g/L

density

2.48 g/cm3 at 20 °C

bulk density

400 kg/m3

anion traces

chloride (Cl-): ≤0.005%
hexafluorosilicate (SiF62-): ≤0.05%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
Na: ≤0.2%
excess alkali: ≤0.05% (as KOH)
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[K+].[F-]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Preanalytical factors influencing the results of ethanol analysis in postmortem specimens.: This study explores various preanalytical factors that affect the accuracy of ethanol analysis in postmortem specimens. Potassium fluoride is highlighted for its role in preventing the postmortem production of ethanol, thus ensuring reliable analytical results (Olds & Jones, 2024).

  • Enantioselective Synthesis of Chiral 2-Nitroallylic Amines via Cooperative Cation-Binding Catalysis.: This study reports the enantioselective synthesis of chiral 2-nitroallylic amines using a cooperative cation-binding catalysis approach involving potassium fluoride. It highlights the compound′s efficacy in asymmetric synthesis, an important area in organic chemistry (Park et al., 2023).

  • Synthesis of Sulfonyl Fluorides Using Direct Chloride/Fluoride Exchange in Potassium Fluoride and Water/Acetone.: The article presents a method for synthesizing sulfonyl fluorides using direct chloride/fluoride exchange in potassium fluoride and water/acetone, highlighting a practical and efficient approach to synthesizing these important compounds in organic chemistry (Bare, 2023).

Analysis Note

Assay (precipitative titration): ≥ 99.0 %
Free alkali (as KOH): ≤ 0.05 %
Free acid (as HF): ≤ 0.05 %
Chloride (Cl): ≤ 0.005 %
Hexafluorosilicate (SiF₆): ≤ 0.05 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Na (Sodium): ≤ 0.2 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service