1.00253
Formic acid
89-91%, for analysis EMSURE® ACS
Synonym(s):
Methanoic acid solution, Formylic acid solution
About This Item
product name
Formic acid 89-91%, for analysis EMSURE® ACS
grade
ACS reagent
Quality Level
product line
EMSURE®
Assay
89.0-91.0% (acidimetric)
form
liquid
autoignition temp.
485 °C
expl. lim.
10-45.5 % (v/v)
impurities
≤0.02 ppm Ag (Silver)
≤0.02 ppm Be (Beryllium)
≤0.02 ppm Co (Cobalt)
≤0.02 ppm Cu (Copper)
≤0.02 ppm Li (Lithium)
≤0.02 ppm Mo (Molybdenum)
≤0.02 ppm Pb (Lead)
≤0.02 ppm Sr (Strontium)
≤0.05 ppm Al (Aluminium)
≤0.05 ppm Ba (Barium)
≤0.05 ppm Cd (Cadmium)
≤0.05 ppm Cr (Chromium)
≤0.05 ppm Ge (Germanium)
≤0.05 ppm Mn (Manganese)
≤0.05 ppm Ni (Nickel)
≤0.05 ppm Tl (Thallium)
≤0.05 ppm V (Vanadium)
≤0.05 ppm Zn (Zinc)
≤0.1 ppm K (Potassium)
≤0.1 ppm Ti (Titanium)
≤0.1 ppm Zr (Zirconium)
≤0.10 ppm Bi (Bismuth)
≤0.2 ppm Ca (Calcium)
≤0.5 ppm Mg (Magnesium)
≤0.5 ppm Na (Sodium)
≤10 ppm NH? (Ammonium)
≤10 ppm Non volatile matter
≤10 ppm Sulfite (SO3)
≤2 ppm Fe (Iron)
≤5 ppm Chloride (Cl)
≤5 ppm Heavy metals (as Pb)
≤5 ppm Sulphate (SO4)
≤500 ppm Acetic acid (CH3COOH)
color
APHA: ≤10
bp
107 °C/1013 hPa
mp
-9 °C
transition temp
flash point 71 °C
density
1.2 g/cm3 at 20 °C
storage temp.
15-25°C
Analysis Note
Colour: ≤ 10 Hazen
Acetic acid (CH₃COOH): ≤ 500 ppm
Chloride (Cl): ≤ 5 ppm
Sulphate (SO₄): ≤ 5 ppm
Sulfite (SO₃): ≤ 10 ppm
Heavy metals (as Pb): ≤ 5 ppm
Ag (Silver): ≤ 0.02 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 0.05 ppm
Be (Beryllium): ≤ 0.02 ppm
Bi (Bismuth): ≤ 0.10 ppm
Ca (Calcium): ≤ 0.2 ppm
Cd (Cadmium): ≤ 0.05 ppm
Co (Cobalt): ≤ 0.02 ppm
Cr (Chromium): ≤ 0.05 ppm
Cu (Copper): ≤ 0.02 ppm
Fe (Iron): ≤ 2 ppm
Ge (Germanium): ≤ 0.05 ppm
K (Potassium): ≤ 0.1 ppm
Li (Lithium): ≤ 0.02 ppm
Mg (Magnesium): ≤ 0.5 ppm
Mn (Manganese): ≤ 0.05 ppm
Mo (Molybdenum): ≤ 0.02 ppm
Na (Sodium): ≤ 0.5 ppm
NH₄ (Ammonium): ≤ 10 ppm
Ni (Nickel): ≤ 0.05 ppm
Pb (Lead): ≤ 0.02 ppm
Sr (Strontium): ≤ 0.02 ppm
Ti (Titanium): ≤ 0.1 ppm
Tl (Thallium): ≤ 0.05 ppm
V (Vanadium): ≤ 0.05 ppm
Zn (Zinc): ≤ 0.05 ppm
Zr (Zirconium): ≤ 0.1 ppm
Non volatile matter: ≤ 10 ppm
Dilution test: passes test
ACS
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
159.8 °F
Flash Point(C)
71 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service