Skip to Content
Merck
All Photos(3)

Key Documents

307491

Sigma-Aldrich

Zinc sulfate monohydrate

≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · H2O
CAS Number:
Molecular Weight:
179.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99.9% trace metals basis
form:
powder
solubility:
water: soluble(lit.)

Quality Level

Assay

≥99.9% trace metals basis

form

powder

solubility

water: soluble(lit.)

SMILES string

O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.H2O.Zn/c1-5(2,3)4;;/h(H2,1,2,3,4);1H2;/q;;+2/p-2

InChI key

RNZCSKGULNFAMC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Fabrication and Characterizations of Hot-Melt Extruded Nanocomposites Based on Zinc Sulfate Monohydrate and Soluplus.: This study details the preparation and characterization of nanocomposites using zinc sulfate monohydrate and Soluplus via hot-melt extrusion. The research highlights the potential applications of these nanocomposites in enhancing the bioavailability of pharmaceutical compounds (Lee et al., 2017).



Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High yield synthesis and characterization of aqueous stable zinc oxide nanocrystals using various precursors.
Swati G, et al.
J. Alloy Compounds, 571, 1-5 (2013)
Jiří Plíšek et al.
Journal of separation science, 36(23), 3702-3708 (2013-10-15)
Biomarkers, 25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 , are important indicators of the vitamin D general status and are monitored in several pathophysiological disorders, such as osteoporosis, diabetes, heart disease, etc. A novel ultra-HPLC with MS/MS methodology for the analysis of
Perry DL.
Handbook of Inorganic Compounds, 457-457 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service