Skip to Content
Merck
All Photos(1)

Key Documents

96495

Sigma-Aldrich

Zinc sulfate monohydrate

purum p.a., ≥99.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · H2O
CAS Number:
Molecular Weight:
179.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (KT)
form:
powder
solubility:
water: soluble(lit.)

grade

purum p.a.

Quality Level

Assay

≥99.0% (KT)

form

powder

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤500 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg

SMILES string

O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.H2O.Zn/c1-5(2,3)4;;/h(H2,1,2,3,4);1H2;/q;;+2/p-2

InChI key

RNZCSKGULNFAMC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Zinc sulfate monohydrate may be used in the following studies:
  • Preparation of rayon, sprays (used in agriculture) and dyes.
  • As a precipitation reagent during the preparation of sample solution for ultra high pressure liquid chromatographic (UHPLC) analysis.
  • Preparation of ZnO nanocrystals, via sol-gel incineration technique.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service