Skip to Content
Merck
All Photos(1)

Key Documents

13440

Sigma-Aldrich

Sodium hypochlorite solution

6-14% active chlorine basis

Synonym(s):

Antiformin, Hypochlorous acid, sodium salt, Sodium chloride oxide (NaClO), Sodium hypochloride, Sodium oxychloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO
CAS Number:
Molecular Weight:
74.44
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Quality Level

shelf life

limited

reaction suitability

reagent type: oxidant

concentration

6-14% (active chlorine)

storage temp.

2-8°C

SMILES string

[Na+].[O-]Cl

InChI

1S/ClO.Na/c1-2;/q-1;+1

InChI key

SUKJFIGYRHOWBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Caution

The assay refers to the freshly manufactured product.
A gradual, substance-specific content reduction, especially at higher temperatures must be expected.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ewa Oleńska et al.
Plants (Basel, Switzerland), 9(8) (2020-08-13)
Heavy metals in soil, as selective agents, can change the structure of plant-associated bacterial communities and their metabolic properties, leading to the selection of the most-adapted strains, which might be useful in phytoremediation. Trifolium repens, a heavy metal excluder, naturally
Heng Zhang et al.
Analytica chimica acta, 1080, 170-177 (2019-08-15)
A sensitive nanocomplex probe prepared from fluorescent polydopamine nanoparticles (F-PDA) and cobalt oxyhydroxide (CoOOH) nanosheets was established for the determination of α-glucosidase activity. In this detection system, the fluorescence of F-PDA was firstly quenched by CoOOH nanosheets based on fluorescence
Bentolhoda Heli et al.
Scientific reports, 10(1), 14367-14367 (2020-09-03)
In this study, we propose a novel sensitive solid-based immunosensor developed on a plasmonic nanopaper platform for the detection of Escherichia coli (E. coli) bacteria. This plasmonic nanopaper that comprises of carboxylated bacterial cellulose (CBC) impregnated with gold nanoparticles (AuNP-CBC)
Aakriti Bhandari et al.
Journal, genetic engineering & biotechnology, 19(1), 40-40 (2021-03-16)
Sophora mollis Royle (family Fabaceae, subfamily-Papilionaceae) is a multipurpose legume distributed in plains and foothills of the North-West Himalaya to Nepal and is facing high risk of extinction due to habitat loss and exploitation by the local people for its
Ina Lefering et al.
Optics express, 26(19), 24384-24402 (2018-11-25)
The nature and magnitude of measurement uncertainties (precision and accuracy) associated with two approaches for measuring absorption by turbid waters (b(532 nm) ranging from 0.20 m-1 to 22.89 m-1) are investigated here: (a) point source integrating cavity absorption meters (PSICAM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service