Skip to Content
Merck
All Photos(5)

Key Documents

217263

Sigma-Aldrich

Sodium thiosulfate

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3
CAS Number:
Molecular Weight:
158.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder, crystals or chunks

grade

reagent

product line

ReagentPlus®

Assay

99%

form

powder, crystals or chunks

pH

6.0-8.5 (20 °C, 50 g/L)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

AKHNMLFCWUSKQB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium thiosulfate is an inorganic salt composed of thiosulfate and sodium ion with the chemical formula Na2S2O3. It is general reducing agent and useful sulfur source in synthetic chemistry

Application

Applied in a widely useful, thiosulfate-assisted, synthesis of a variety of diamine-CuCN complexes.
Sodium thiosulfate can be used as a
  • Component in the synthesis of unsymmetrical aryl sulfides under aqueous conditions, where C–S–C bond formed in presence of other two components (alcohols and organohalides).
  • Reagent for the activation of carbon material.
  • Reagent in the synthesis of S-glycosyl thiosulfates (glycosyl bunte salts) by direct condensation of unprotected sugars using dehydrating agent DMC (2-chloro-1,3-dimethylimidazolinium chloride) in water-acetonitrile solvent mixture.
  • Reagent with alkyl halide in the synthesis of Bunte salts by SN2 mechanism using Cu catalyst.
  • Reagent in the synthesis of aryl thiols from aryl halides by palladium catalyzed cross-coupling reaction.

Features and Benefits

Sodium thiosulfate is:
  • Stable
  • Environmentally friendly
  • Odorless
  • Inexpensive.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 648-648 (1994)
Xin Yan Hou et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 125, 285-289 (2014-02-22)
In this study, triangular silver nanoplates (TAg-NPs) were used to detect trace Cysteine concentration in the presence of sodium thiosulfate (Na2S2O3). Study showed that the TAg-NPs could be gently etched by Cysteine with the concentration of 1.0×10(-7) mol L(-1) through
Eagleson M.
Concise Encyclopedia Chemistry, 542-542 (1994)
Hongda Wang et al.
Light, science & applications, 9, 118-118 (2020-07-21)
Early identification of pathogenic bacteria in food, water, and bodily fluids is very important and yet challenging, owing to sample complexities and large sample volumes that need to be rapidly screened. Existing screening methods based on plate counting or molecular
Fred B. Stocker et al.
Inorganic chemistry, 38(5), 984-991 (2001-10-24)
A new synthetic procedure developed recently in our laboratories has made possible the synthesis of variety of new complexes of CuCN with diamines. Synthesis was effected by adding the ligand to a solution of CuCN in aqueous sodium thiosulfate. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service