Skip to Content
Merck
All Photos(1)

Key Documents

Y0001287

Heparin calcium for NMR identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Heparin calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

heparin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Heparin calcium for NMR identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Andreozzi et al.
Minerva cardioangiologica, 48(12 Suppl 1), 9-14 (2001-03-20)
Thrombophlebitis of the superficial veins (SVT) of the leg is usually regarded as a mild and uncomplicated disease. Although this is generally true for acute thrombosis of the branches of the saphenous vein, the natural history of SVT involving the
C E Geffroy et al.
Presse medicale (Paris, France : 1983), 31(7), 303-311 (2002-03-20)
To control whether prescriptions of curative doses of heparin (non fractioned heparins, enoxaparin, tinzaparin) in the hospital complied with the official recommendations; to provide the physicians with information adapted to the recorded misuse and to evaluate the influence of this
Cutaneous allergic reaction to heparins: subcutaneous but not intravenous provocation.
Maria Maroto-Iitani et al.
Contact dermatitis, 52(4), 228-230 (2005-04-30)
Maria Laura Cossu et al.
Chirurgia italiana, 59(3), 331-335 (2007-08-01)
Patients with morbid obesity who undergo bariatric surgery are usually considered at high risk of developing venous thromboembolism. Considering that deep vein thrombosis is often asymptomatic, primary prevention is the key to reducing morbidity and mortality. Between 1995 and 2003
F Perfetto et al.
Annali italiani di medicina interna : organo ufficiale della Societa italiana di medicina interna, 16(3), 179-184 (2001-11-06)
An 80-year-old woman was admitted to the Traumatologic Hospital of Florence because of bilateral post-traumatic humeral fractures. Subcutaneous calcium heparin was immediately administered to prevent deep venous thrombosis. Nine days later, the patient was referred to the Internal Medicine Unit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service