Skip to Content
Merck
All Photos(1)

Key Documents

H4784

Sigma-Aldrich

Heparin sodium salt from porcine intestinal mucosa

≥180 USP units/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

biological source

Porcine intestinal mucosa

Quality Level

form

powder

specific activity

≥180 USP units/mg

solubility

H2O: 50 mg/mL, clear, colorless to faintly yellow

cation traces

Ca: ≤20 ppm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Quality

Low calcium content

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Ye et al.
PloS one, 8(1), e54622-e54622 (2013-01-30)
The application of polyelectrolyte multilayer films is a new, versatile approach to surface modification of decellularized tissue, which has the potential to greatly enhance the functionality of engineered tissue constructs derived from decellularized organs. In the present study, we test
Eve Draper et al.
PloS one, 8(5), e61825-e61825 (2013-05-08)
The current generation of Human Papillomavirus (HPV) vaccines, Cervarix® and Gardasil®, exhibit a high degree of efficacy in clinical trials against the two high-risk (HR) genotypes represented in the vaccines (HPV16 and HPV18). High levels of neutralizing antibodies are elicited
Linda Cruz et al.
PloS one, 8(7), e68379-e68379 (2013-07-19)
Human papillomavirus (HPV) infection is the leading cause of cervical cancer world-wide. Here, we show that native HPV particles produced in a differentiated epithelium have developed different strategies to infect the host. Using biochemical inhibition assays and glycosaminoglycan (GAG)-negative cells
William H Thiel et al.
PloS one, 7(9), e43836-e43836 (2012-09-11)
The broad applicability of RNA aptamers as cell-specific delivery tools for therapeutic reagents depends on the ability to identify aptamer sequences that selectively access the cytoplasm of distinct cell types. Towards this end, we have developed a novel approach that
Elena Garreta et al.
Physiological reports, 2(7) (2014-10-28)
Whole-organ decellularization technology has emerged as a new alternative for the fabrication of bioartificial lungs. Embryonic stem cells (ESC) and induced pluripotent stem cells (iPSC) are potentially useful for recellularization since they can be directed to express phenotypic marker genes

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service