Skip to Content
Merck
All Photos(1)

Key Documents

Y0001259

Enrofloxacin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Baytril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O3
CAS Number:
Molecular Weight:
359.39
Beilstein:
5307824
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

enrofloxacin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN1CCN(CC1)c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)C4CC4

InChI

1S/C19H22FN3O3/c1-2-21-5-7-22(8-6-21)17-10-16-13(9-15(17)20)18(24)14(19(25)26)11-23(16)12-3-4-12/h9-12H,2-8H2,1H3,(H,25,26)

InChI key

SPFYMRJSYKOXGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enrofloxacin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

María-Ysabel Piñero et al.
Electrophoresis, 33(19-20), 2978-2986 (2012-09-29)
An easy, selective, and sensitive method has been developed for the determination of enrofloxacin (ENR) and its main active metabolite, ciprofloxacin (CIP), in raw bovine milk using CE with UV detection at 268 nm. Milk samples were prepared by a
Disposition of single-dose oral enrofloxacin in the horse.
V C Langston et al.
Journal of veterinary pharmacology and therapeutics, 19(4), 316-319 (1996-08-01)
Kristine H Wammer et al.
Water research, 47(1), 439-448 (2012-11-13)
Fluoroquinolone (FQ) antibacterial compounds are frequently detected in the aquatic environment, and photodegradation is expected to play an important role in FQ fate in some sunlit surface waters. This study investigated the direct aquatic photochemistry of three FQs: norfloxacin, ofloxacin
Jochen Springer et al.
Stroke, 45(12), 3675-3683 (2014-10-30)
Muscle wasting is a common complication accompanying stroke. Although it is known to impair poststroke recovery, the mechanisms of subacute catabolism after stroke have not been investigated in detail. The aim of this study is to investigate mechanisms of local
Sook Mei Khor et al.
Analytical and bioanalytical chemistry, 405(11), 3889-3898 (2013-02-27)
A displacement immunoassay involves having a labelled analogue of the analyte (the epitope) already bound to the antibody. The presence of the analyte causes a competition for antibodies, and some of the antibodies dissociates from the epitope so that it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service