Skip to Content
Merck
All Photos(1)

Key Documents

17849

Sigma-Aldrich

Enrofloxacin

≥99.0%

Synonym(s):

Baytril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O3
CAS Number:
Molecular Weight:
359.39
Beilstein:
5307824
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0%
99.0-101.0% (dried substance)

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

CCN1CCN(CC1)c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)C4CC4

InChI

1S/C19H22FN3O3/c1-2-21-5-7-22(8-6-21)17-10-16-13(9-15(17)20)18(24)14(19(25)26)11-23(16)12-3-4-12/h9-12H,2-8H2,1H3,(H,25,26)

InChI key

SPFYMRJSYKOXGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: fluoroquinolone
Enrofloxacin is a veterinary fluoroquinolone (FQ) antibiotic with broad-spectrum antimicrobial activity. FQ drugs chelate to iron with high affinity. This chelation leads to inhibition of Fe(II)-dependent dioxygenases involved in the regulation of epigenetic control, collagen maturation, and HIF (hypoxia-inducible factor) degradation. The side effects associated with FQ drugs are renal toxicity and tendinopathy. Enrofloxacin is used in aquaculture and its use may cause an increase in the growth of antibiotic resistant microbes in an aquatic environment. The drug may have negative effects on public health and environment.[1][2][3]

Application

Enrofloxacin has been used:
  • to investigate the pathological mechanisms resulting from the toxicity of fluoroquinolones in mammalian cells[1]
  • to prepare a standard solution with 50% acetonitrile in a study to analyze illegal fish drugs used in aquaculture by employing surface-enhanced Raman spectroscopy (SERS)[2]
  • as an analyte in a chemiluminescence reagent system[4]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica M Terry et al.
The Analyst, 136(5), 913-919 (2010-12-04)
Constructing flow-through reactors for chemiluminescence detection by machining channels into polymer disks has enabled the exploration of new configurations and materials that can improve signal intensity beyond that attainable with the traditional coiled-tubing design. Several approaches to merge reactant solutions
Yanina N Martínez et al.
Bioresource technology, 145, 280-284 (2013-04-06)
A keratinase isolated from Paecilomyces lilacinus (LPS #876) was tested against proteins present in the skin but the high enzyme activity was detected on collagen. Keratinase was physically immobilized onto PVA-pectin cryogels and enzyme release was 20.8±2.1%, 63.8±0.2%, 41.5±3.5% and
Geaneth Pertunia Mashile et al.
Polymers, 12(5) (2020-05-16)
The application of a magnetic mesoporous carbon/β-cyclodextrin-chitosan (MMPC/Cyc-Chit) nanocomposite for the adsorptive removal of danofloxacin (DANO), enrofloxacin (ENRO) and levofloxacin (LEVO) from aqueous and environmental samples is reported in this study. The morphology and surface characteristics of the magnetic nanocomposite
Susanne Rath et al.
Chemosphere, 214, 111-122 (2018-09-28)
Brazil is one of the world's largest producers of animal protein, requiring the large-scale use of veterinary drugs. The administration of antimicrobials and antiparasitics is a common practice. However, there is a lack of information on how these drugs impact
Kristine H Wammer et al.
Water research, 47(1), 439-448 (2012-11-13)
Fluoroquinolone (FQ) antibacterial compounds are frequently detected in the aquatic environment, and photodegradation is expected to play an important role in FQ fate in some sunlit surface waters. This study investigated the direct aquatic photochemistry of three FQs: norfloxacin, ofloxacin

Questions

1–2 of 2 Questions  
  1. Is it possible to store the drug frozen for long-term preservation after dissolving it in organic solvents?

    1 answer
    1. The solution stability has not been tested. However, literature research suggests that enrofloxacin solutions dissolved in organic solvents such as methanol, ethanol, and dimethyl sulfoxide (DMSO) stored under frozen conditions can typically remain stable for approximately 26 to 52 weeks (6 to 12 months). The exact stability can depend on factors such as concentration, formulation, and specific storage conditions.

      Helpful?

  2. What is the solubility of Antibiotic Question 17849 in water? This is intended for use in a mouse experiment involving drinking water.

    1 answer
    1. The substance is not soluble in water. It has a solubility of 1mg/ml in organic solvents such as DMSO and 10 mg/ml in DMF. Additionally, it is suggested to try if the drug dissolves in (2-Hydroxypropyl)-β-cyclodextrin solution H5784.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service