Skip to Content
Merck
All Photos(1)

Key Documents

Y0000659

Pyrrolidone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Pyrrolidinone, 2-Pyrrolidone, Butyrolactam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO
CAS Number:
Molecular Weight:
85.10
Beilstein:
105241
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

vapor density

2.9 (vs air)

API family

levetiracetam, piracetam

manufacturer/tradename

EDQM

refractive index

n20/D 1.487 (lit.)

bp

245 °C (lit.)

mp

23-25 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1CCCN1

InChI

1S/C4H7NO/c6-4-2-1-3-5-4/h1-3H2,(H,5,6)

InChI key

HNJBEVLQSNELDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pyrrolidone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srimanta Sarkar et al.
International journal of pharmaceutics, 475(1-2), 558-565 (2014-09-15)
This study investigated the influence of different disintegrants, present in different substrate physical forms, on dimensional recovery of multi-component tablets prepared at different compression pressures. Formulations containing model drug, metformin, (10%, w/w), different disintegrants (10%, w/w), and lactose (80%, w/w)
Jani Holopainen et al.
Nanotechnology, 26(2), 025301-025301 (2014-12-17)
A needleless electrospinning setup named 'Needleless Twisted Wire Electrospinning' was developed. The polymer solution is electrospun from the surface of a twisted wire set to a high voltage and collected on a cylindrical collector around the wire. Multiple Taylor cones
J Alex Grizzell et al.
Behavioural brain research, 268, 55-65 (2014-04-10)
Chronic stress underlies and/or exacerbates many psychiatric conditions and often results in memory impairment as well as depressive symptoms. Such afflicted individuals use tobacco more than the general population and this has been suggested as a form of self-medication. Cotinine
A S Chudasama et al.
Indian journal of pharmaceutical sciences, 76(3), 218-224 (2014-07-19)
Nevirapine is a highly lipophilic and water insoluble non-nucleoside reverse transcriptase inhibitor used for the treatment of HIV-1 infection. Lymphoid tissue constitutes the major reservoir of HIV virus and infected cells in HIV-infected patients. Self-emulsifying drug delivery system, using long
Norhafiza Razali et al.
Clinical & experimental ophthalmology, 43(1), 54-66 (2014-07-06)
Steroid-induced ocular hypertension is currently treated in the same way as primary open-angle glaucoma. However, the treatment is often suboptimal and is associated with adverse effects. We evaluated the oculohypotensive effects of topical trans-resveratrol in rats with steroid-induced ocular hypertension

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service