Skip to Content
Merck
All Photos(4)

Key Documents

H5501

Sigma-Aldrich

Methyl 4-hydroxybenzoate

ReagentPlus®, ≥99.0%, crystalline

Synonym(s):

p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
509801
EC Number:
MDL number:
UNSPSC Code:
51452201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

crystalline

mp

125-128 °C (lit.)

SMILES string

COC(=O)c1ccc(O)cc1

InChI

1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3

InChI key

LXCFILQKKLGQFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 4-hydroxybenzoate has been used:
  • in sex-linked recessive lethal (SLRL) test in Drosophila melanogaster
  • as a constituent in cream formulation
  • as an antifungal agent in Drosophila melanogaster culture

Biochem/physiol Actions

Methyl 4-hydroxybenzoate, also called methyl paraben or nipagin, comprises the ester of p-hydroxybenzoic acid. It is present naturally in cloudberry, white wine and bourbon vanilla. Methyl 4-hydroxybenzoate has antimicrobial and antifungal functionality and is commercially used as a preservative in the food, cosmetic and pharmaceutical industry. Methyl 4-hydroxybenzoate has cytotoxic effects on keratinocytes in the presence of sunlight. Methyl 4-hydroxybenzoate upon solar irradiation mediates DNA damage and modulates esterase metabolism resulting in skin damage and favors cancer progression. Methyl 4-hydroxybenzoate has estrogenic functionality and upregulates estrogen-related genes.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fruit flies may face a nutrient-dependent life-history trade-off between secondary sexual trait quality, survival and developmental rate
Gray LJ, et al.
Journal of Insect Physiology, 104(8), 60-70 (2018)
Determination of methylparaben from cosmetic products by ultra performance liquid chromatography.
Mincea M, et al.
J. Serb. Chem. Soc., 74(6), 1335-1373 (2009)
Combined activation of methyl paraben by light irradiation and esterase metabolism toward oxidative DNA damage
Okamoto Y, et al.
Chemical Research in Toxicology, 21(8), 1594-1599 (2008)
The estrogenicity of methylparaben and ethylparaben at doses close to the acceptable daily intake in immature Sprague-Dawley rats
Sun L, et al.
Scientific Reports, 6, 25173-25173 (2016)
Simultaneous Determination of Fusidic Acid, m-and p-Hydroxybenzoates and Butylhydroxyanisole by TLC with Densitometric Detection in UV
Krzek J, et al.
Journal of Liquid Chromatography and Related Technologies, 29(14), 2129-2139 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service