Skip to Content
Merck
All Photos(1)

Key Documents

Y0000507

Chlorpromazine impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Chlorpromazine sulfoxide, 3-(2-Chloro-10H-phenothiazin-10-yl)-N,N-dimethylpropan-1- amine S-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19ClN2OS
CAS Number:
Molecular Weight:
334.86
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlorpromazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C17H19ClN2OS/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)22(21)17-9-8-13(18)12-15(17)20/h3-4,6-9,12H,5,10-11H2,1-2H3

InChI key

QEPPAOXKZOTMPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorpromazine impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Nakano et al.
Biochemical and biophysical research communications, 130(3), 952-956 (1985-08-15)
Chemiluminescence in visible region was detected, during peroxidase-catalyzed oxidation of chlorpromazine at pH7.5 (but not at pH4.25) or of propericiazine, both at pH7.5 and at pH4.25. Red colored intermediates, cation radicals, were produced and decayed in all enzymatic systems used.
Free radical production by chlorpromazine sulfoxide, an ESR spin-trapping and flash photolysis study.
G R Buettner et al.
Photochemistry and photobiology, 44(1), 5-10 (1986-07-01)
Z Y Sahaf et al.
Brain research, 437(2), 397-401 (1987-12-29)
Treatment of frog neuromuscular preparations with chlorpromazine (5 mumol/l) resulted in a marked rise in miniature endplate potential (MEPP) frequency of greater than 100% within 30 min, and an increase in evoked transmitter release (quantal content 5-15) of about 35%.
P K Yeung et al.
The Journal of pharmacology and experimental therapeutics, 226(3), 833-838 (1983-09-01)
Antibody specific for chlorpromazine sulfoxide (CPZSO) was produced in rabbits immunized with a hapten-bovine serum albumin conjugate, which was prepared by linking the 10-alkyl side chain of CPZSO to the protein molecule via a 2-carbon bridge. A simple radioimmunoassay was
S G Dahl et al.
Molecular pharmacology, 21(2), 409-414 (1982-03-01)
The 3-dimensional molecular structures of methotrimeprazine, methotrimeprazine sulfoxide, and chlorpromazine sulfoxide were examined by X-ray crystallography. Previous studies of their dopamine receptor binding affinities have indicated that both chlorpromazine sulfoxide and methotrimeprazine sulfoxide lack neuroleptic potency. The crystal structures of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service