Skip to Content
Merck
All Photos(1)

Key Documents

Y0000479

Chlorpromazine impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Desmethylchlorpromazine, 3-(2-Chloro-10H-phenothiazin-10-yl)-N-methylpropan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17ClN2S
CAS Number:
Molecular Weight:
304.84
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chlorpromazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C16H17ClN2S/c1-18-9-4-10-19-13-5-2-3-6-15(13)20-16-8-7-12(17)11-14(16)19/h2-3,5-8,11,18H,4,9-10H2,1H3

InChI key

YHFXGBOUSIKGMZ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorpromazine impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Alfredsson et al.
Acta psychiatrica Scandinavica. Supplementum, 311, 49-74 (1984-01-01)
Schizophrenic patients were treated with fixed doses of sulpiride (800 mg) or chlorpromazine (400 mg) during eight weeks using a double-blind design. In order to examine relationships between pharmacokinetic, clinical and biochemical parameters in relation to treatment the following variables
M Chetty et al.
Therapeutic drug monitoring, 13(4), 350-355 (1991-07-01)
The concentrations of chlorpromazine (CPZ) and six of its metabolites in patient plasma samples that had been stored for 24 h at -20 degrees C, for 1 week at -20 degrees C, and for 4 weeks at -70 degrees C
D L Newton et al.
Biochemistry, 28(9), 3750-3757 (1989-05-02)
In the presence of Ca2+ norchlorpromazine isothiocyanate forms a monocovalent complex with calmodulin: CAPP1-calmodulin (Newton et al, 1983). Trypsin digestion of [3H]CAPP1-calmodulin yields as the major radioactive peptide N epsilon-CAPP-Lys-Met-Lys, corresponding to residues 75-77 of calmodulin. Stoichiometric amounts of all
G P Sgaragli et al.
Bollettino della Societa italiana di biologia sperimentale, 60(9), 1757-1763 (1984-09-30)
Nanogram amounts of chlorimipramine, chlorpromazine and their Nor1- and Nor2-metabolites were detected in plasma by GLC with nitrogen-sensitive detection. Two extraction procedures were compared. The use of Sep Pak C18 cartridges produced a higher degree of accuracy and precision with
R Ninci et al.
Journal of chromatography, 381(2), 315-322 (1986-09-05)
A gas chromatographic method using nitrogen-selective detection for the quantitative determination of nanogram amounts of chlorimipramine, chlorpromazine and their nor1 and nor2 derivatives in plasma is described. Derivatization with trifluoroacetic anhydride of nor1 and nor2 metabolites allowed the chromatographic separation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service