Skip to Content
Merck
All Photos(1)

Key Documents

Y0000468

Brotizolam

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10BrClN4S
CAS Number:
Molecular Weight:
393.69
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

brotizolam

manufacturer/tradename

EDQM

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C15H10BrClN4S/c1-8-19-20-13-7-18-14(9-4-2-3-5-11(9)17)10-6-12(16)22-15(10)21(8)13/h2-6H,7H2,1H3

InChI key

UMSGKTJDUHERQW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Brotizolam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryoko Kawahara et al.
Masui. The Japanese journal of anesthesiology, 54(11), 1263-1267 (2005-11-22)
Hypnotics have been used to provide adequate sleep on the night before surgery, but these effects have rarely been investigated. We investigated whether the preoperative hypnotic effects are related with the severity of surgery with quantitative evaluation using actigraph. Sleep
Hiromi Katoh et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 127(12), 2035-2044 (2007-12-07)
In the present study, we tested three kinds of sleeping drugs, consisting mainly of triazolam, brotizolam, and flunitrazepam, to compare the drug efficacy of generic drugs with that of original drugs. After these drugs were administered orally to mice, drug
M Tanaka et al.
Methods and findings in experimental and clinical pharmacology, 30(8), 607-613 (2008-12-18)
In this study we examined the behavioral pharmacological side effects after recovery from the loss of righting reflex induced by three benzodiazepine receptor agonists - zolpidem, brotizolam and flunitrazepam - in ddY mice. All agents caused marked motor incoordination in
Akshay Anand et al.
Behavioural brain research, 182(1), 12-20 (2007-06-15)
Benzodiazepines such as diazepam, lorazepam, are reported to produce anterograde amnesia but these do not affect the retrieval mechanism. Triazodiazepines such as alprazolam, triazolam and brotizolam produce both anterograde and retrograde amnesia. Because benzodiazepine receptor antagonists are known to reverse
Yukihiro Ujiie et al.
Therapeutic drug monitoring, 28(3), 299-302 (2006-06-17)
The purpose of the present study was to examine the effects of rifampicin on the single oral dose pharmacokinetics and pharmacodynamics of brotizolam. Thirteen healthy male volunteers received rifampicin 450 mg/day, or matched placebo, for 7 days in a double-blind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service