Skip to Content
Merck
All Photos(1)

Key Documents

Y0000090

Finasteride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(2-methyl-2-propyl)-3-oxo-4-aza-5α-androst-1-ene-17β-carboxamide, N-tert-Butyl-3-oxo-4-aza-5α-androst-1-en-17β-carboxamide, MK-906

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36N2O2
CAS Number:
Molecular Weight:
372.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

finasteride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(=O)NC(C)(C)C)[C@@]4(C)CC[C@]3([H])[C@@]1(C)C=CC(=O)N2

InChI

1S/C23H36N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h11,13-18H,6-10,12H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1

InChI key

DBEPLOCGEIEOCV-WSBQPABSSA-N

Gene Information

human ... SRD5A2(6716)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Finasteride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Selective 5α-reductase inhibitor; antiandrogen.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Jin Li et al.
Cancer prevention research (Philadelphia, Pa.), 2(6), 518-524 (2009-06-06)
Our inability to distinguish between low-grade prostate cancers that pose no threat and those that can kill compels newly diagnosed early prostate cancer patients to make decisions that may negatively affect their lives needlessly for years afterward. To reliably stratify
Cosimo De Nunzio et al.
Expert opinion on drug metabolism & toxicology, 4(12), 1561-1568 (2008-12-02)
The development and introduction in clinical practice of finasteride, the first potent inhibitor of steroid 5-alpha-reductase that inhibits the conversion of testosterone to its active form dihydroxytestosterone, has created the possibility to prevent and treat prostatic disorder by manipulating and
Sarah L Hulin-Curtis et al.
Future oncology (London, England), 6(12), 1897-1913 (2010-12-15)
Incidences of prostate cancer in most countries are increasing owing to better detection methods; however, prevention with the use of finasteride, a very effective steroid 5α-reductase type II inhibitor, has been met with mixed success. A wide interindividual variation in
Manlio A Goetzl et al.
Nature clinical practice. Urology, 3(8), 422-429 (2006-08-12)
Prostate cancer chemoprevention involves the use of natural and/or synthetic agents that inhibit or reverse the development of precancerous lesions or delay progression of these lesions to invasive disease. The recent completion of the first Phase III trial for prostate
Sergio Vañó-Galván et al.
Journal of the American Academy of Dermatology, 70(4), 670-678 (2014-02-11)
To our knowledge, there are no large multicenter studies concerning frontal fibrosing alopecia (FFA) that could give clues about its pathogenesis and best treatment. We sought to describe the epidemiology, comorbidities, clinical presentation, diagnostic findings, and therapeutic choices in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service