Skip to Content
Merck
All Photos(1)

Key Documents

Y0000040

Ciclopirox

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ciclopirox

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

ON1C(C2CCCCC2)=CC(C)=CC1=O

InChI

1S/C12H17NO2/c1-9-7-11(13(15)12(14)8-9)10-5-3-2-4-6-10/h7-8,10,15H,2-6H2,1H3

InChI key

SCKYRAXSEDYPSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciclopirox EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aditya K Gupta et al.
International journal of dermatology, 45(1), 66-69 (2006-01-24)
Seborrheic dermatitis is a chronic superficial fungal infection of the skin, particularly affecting sites rich in sebaceous glands. Although the precise etiology of seborrheic dermatitis is uncertain, yeasts of the genus Malassezia are known to play a causative role. Ciclopirox
A K Gupta et al.
Journal of the American Academy of Dermatology, 43(4 Suppl), S96-102 (2000-10-29)
Ciclopirox nail lacquer solution 8% has been shown to be effective in the treatment of dermatophyte onychomycosis of mild to moderate severity Other studies report the effectiveness of ciclopirox nail lacquer in onychomycosis caused by Candida sp and nondermatophyte molds.
M Bohn et al.
Journal of the American Academy of Dermatology, 43(4 Suppl), S57-S69 (2000-10-29)
Ciclopirox is a synthetic hydroxypyridone antifungal agent. In contrast to the azoles, glucuronidation is the main metabolic pathway of ciclopirox; therefore interactions with drugs metabolized via the cytochrome P450 system are unlikely Ciclopirox is also distinct from the common systemic
Alessandro Subissi et al.
Drugs, 70(16), 2133-2152 (2010-10-23)
Ciclopirox is a topical antimycotic agent belonging to the chemical class of hydroxypyridones and not related to azoles or any other class of antifungal agents. Its antimicrobial profile includes nearly all of the clinically relevant dermatophytes, yeasts and moulds, and
Ciclopirox: an overview.
A K Gupta
International journal of dermatology, 40(5), 305-310 (2001-09-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service