Skip to Content
Merck
All Photos(1)

Key Documents

17799

Supelco

Rhamnetin

analytical standard

Synonym(s):

β-Rhamnocitrin, 3,3′,4′,5-Tetrahydroxy 7-methoxyflavone, 7-Methoxyquercetin, 7-Methylquercetin, Quercetin 7-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O7
CAS Number:
Molecular Weight:
316.26
Beilstein:
47741
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(O)c2C(=O)C(O)=C(Oc2c1)c3ccc(O)c(O)c3

InChI

1S/C16H12O7/c1-22-8-5-11(19)13-12(6-8)23-16(15(21)14(13)20)7-2-3-9(17)10(18)4-7/h2-6,17-19,21H,1H3

InChI key

JGUZGNYPMHHYRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Schieber et al.
Journal of agricultural and food chemistry, 51(17), 5006-5011 (2003-08-09)
Flavonol O- and xanthone C-glycosides were extracted from mango (Mangifera indica L. cv. "Tommy Atkins") peels and characterized by high-performance liquid chromatography-electrospray ionization mass spectrometry. Among the fourteen compounds analyzed, seven quercetin O-glycosides, one kaempferol O-glycoside, and four xanthone C-glycosides
Leena Pohjala et al.
Molecules (Basel, Switzerland), 17(9), 10774-10790 (2012-09-11)
Previous descriptions of quercetin, a widely studied flavonoid, as a frequently reported nonspecific screening hit due to aggregating behavior has raised questions about the reliability of in vitro bioactivity reports of phenolic compounds. Here a systematic study on 117 phenolic
F Bucar et al.
Die Pharmazie, 53(12), 875-878 (1999-01-08)
From the methylene chloride extract of the leaves of Guiera senegalensis a novel naphthopyran, 5-methylflavasperone (5,8,10-trimethoxy-2-methyl-4 H-naphtho[1,2-b]pyran-4-one), as well as rhamnetin were isolated. Rhamnetin showed a strong inhibitory activity on 5-lipoxygenase from porcine leucocytes with an IC50 value of 0.7
Sunhee Lee et al.
Bioorganic & medicinal chemistry letters, 21(13), 3866-3870 (2011-06-10)
To produce rhamnetin using enzymatic engineering, poplar O-methyltransferase-7 and its mutants were prepared based on the rational enzyme design, and the production of rhamnetin was compared with the results obtained using the wild type enzyme. In addition, the potential of
Andrea Mattarei et al.
Molecules (Basel, Switzerland), 15(7), 4722-4736 (2010-07-27)
The regioselective synthesis of several quercetin (3,3',4',5,7-pentahydroxy flavone) tetraesters bearing a single free OH on 5-C was achieved in good yield by proper choice of reaction conditions using common esterification procedures. Tetracetylated quercetin with the free OH on 7-C was

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service