Skip to Content
Merck
All Photos(1)

Key Documents

W387520

Sigma-Aldrich

Methyl sulfoxide

≥99%, FG

Synonym(s):

Dimethyl sulfoxide, DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
FEMA Number:
3875
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.175
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.859

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; garlic; mushroom; oily; cheesy

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Dimethyl Sulfoxide (DMSO) as a Potential Source of Interference in Research Related to Sulfur Metabolism-A Preliminary Study.: This study investigates the potential interference of DMSO in sulfur metabolism research, highlighting its impact on experimental outcomes. (Kaczor-Kamińska et al., 2024).

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allison L Brill et al.
Scientific reports, 10(1), 386-386 (2020-01-17)
Polycystin 2 (PC2 or TRPP1, formerly TRPP2) is a calcium-permeant Transient Receptor Potential (TRP) cation channel expressed primarily on the endoplasmic reticulum (ER) membrane and primary cilia of all cell and tissue types. Despite its ubiquitous expression throughout the body
J I Bertucci et al.
Aquatic toxicology (Amsterdam, Netherlands), 225, 105523-105523 (2020-06-13)
Hexabromocyclododecane (HBCD) is a ubiquitous environmental contaminant of current concern despite its global ban in 2013 due to its characteristics as a persistent organic pollutant. While the toxicity of HBDC in vertebrates has been extensively studied, the specific molecular mechanisms
Gloria Pérez-Rubio et al.
Diagnostics (Basel, Switzerland), 10(1) (2020-01-08)
Cigarette smoking is influenced by nicotine's effects on dopaminergic activity, which appear to be moderated by genetic variation, particularly a variable number tandem repeat (VNTR, 48 bp) polymorphism in the third exon of the dopamine receptor gene (DRD4). Smokers with
Cefan Zhou et al.
Autophagy, 16(10), 1786-1806 (2019-11-08)
Macroautophagy/autophagy plays key roles in development, oncogenesis, and cardiovascular and metabolic diseases. Autophagy-specific class III phosphatidylinositol 3-kinase complex I (PtdIns3K-C1) is essential for autophagosome formation. However, the regulation of this complex formation requires further investigation. Here, we discovered that STYK1
Cefan Zhou et al.
Molecular cancer, 19(1), 118-118 (2020-07-31)
Pancreatic cancer is one of the most lethal malignancies and has an extremely poor diagnosis and prognosis. The development of resistance to gemcitabine is still a major challenge. The long noncoding RNA PVT1 was reported to be involved in carcinogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service