1.16743
Dimethyl sulfoxide
EMPLURA®
Synonym(s):
Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO, DMSO
Select a Size
Select a Size
About This Item
Recommended Products
vapor density
2.7 (vs air)
Quality Level
vapor pressure
0.42 mmHg ( 20 °C)
product line
EMPLURA®
Assay
≥99% (GC)
form
liquid
autoignition temp.
573 °F
potency
28300 mg/kg LD50, oral (Rat)
40000 mg/kg LD50, skin (Rat)
expl. lim.
42 %, 63 °F
impurities
≤0.001% Free acid (as CH3COOH)
≤0.2% Water
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- A simple and efficient synthesis of 5-hydroxymethylfurfural from carbohydrates using acidic ionic liquid grafted on silica gel.: This study presents a novel method for synthesizing 5-hydroxymethylfurfural, where DMSO plays a critical role in enhancing the efficiency and selectivity of the reaction under mild conditions (Doan VTC et al., 2024).
- Controlling Tin Halide Perovskite Oxidation Dynamics in Solution for Perovskite Optoelectronic Devices.: This paper discusses the role of DMSO in managing oxidation processes in tin halide perovskites, crucial for developing stable and efficient optoelectronic devices (Nazeeruddin MKK et al., 2024).
- Preparation technology comparison and performance evaluation of different protein A affinity chromatographic materials.: Demonstrates the effectiveness of DMSO in optimizing the preparation and performance of chromatographic materials used in protein purification (Zhou LJ et al., 2024).
- Application of copper (I) selective ligands for PET imaging of reactive oxygen species through metabolic trapping.: Highlights a novel application of DMSO in the development of copper (I) selective ligands for positron emission tomography (PET), enhancing imaging techniques used in medical diagnostics (Tada T et al., 2024).
Caution
Specifications
Analysis Note
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.001 %
Density (d 20 °C/ 4 °C): 1.099 - 1.101
Refractive index (n 20/D): 1.478 - 1.479
Melting point: ≥ 18.0 °C
Absorption: conforms
Related substances (GC): conforms
Readily carbonizable substances: conforms
Water: ≤ 0.2 %
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service