Skip to Content
Merck
All Photos(3)

Key Documents

V8138

Sigma-Aldrich

Vancomycin hydrochloride from Streptomyces orientalis

meets USP testing specifications

Synonym(s):

Vancomycin, Vancomycin HCL, Vancomycin Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · HCl
CAS Number:
Molecular Weight:
1485.71
Beilstein:
3704657
UNSPSC Code:
51282703
PubChem Substance ID:
NACRES:
NA.76

biological source

Streptomyces orientalis

Quality Level

Agency

USP/NF
meets USP testing specifications

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

concentration

≥900 μg/mg (as vancomycin base)

antibiotic activity spectrum

Gram-positive bacteria

application(s)

pharmaceutical (small molecule)

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl[H].CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]1O[C@H]2C[C@](C)(N)[C@@H](O)[C@@H](C)O2.CN[C@H](CC(C)C)C(=O)NC3[C@H](O)c4ccc(Oc5cc6Oc7ccc(cc7Cl)[C@@H](O)[C@H]8NC(=O)[C@H](NC(=O)[C@H](NC(=O)[C@H](CC(N)=O)NC3=O)c(c5)c6)c9ccc(O)c(c9)-c%10c(O)cc(O)cc%10[C@@H](NC8=O)C(O)=O)c(Cl)c4

InChI

1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34+,35-,42+,44-,46+,47+,48-,49+,50-,51+,52+,53+,54-,56+,57+,65-,66-;/m0./s1

InChI key

LCTORFDMHNKUSG-XTTLPDOESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: glycopeptide

Biochem/physiol Actions

Mode of action: Inhibits incorporation of terminal D-alanyl-D-alanine moieties of the NAM/NAG-peptides.

Antimicrobial Spectrum: Active against Gram-positive bacteria

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacqueline C Y Lai et al.
Antimicrobial agents and chemotherapy, 64(3) (2019-12-11)
Infection is correlated with increased risk of neurodevelopmental sequelae in preterm infants. In modeling neonatal brain injury, Toll-like receptor agonists have often been used to mimic infections and induce inflammation. Using the most common cause of bacteremia in preterm infants
David W Eyre et al.
The Journal of infectious diseases, 209(9), 1446-1451 (2013-11-13)
Whole-genome sequencing was used to determine whether the reductions in recurrence of Clostridium difficile infection observed with fidaxomicin in pivotal phase 3 trials occurred by preventing relapse of the same infection, by preventing reinfection with a new strain, or by
Wessam Abdelhady et al.
The Journal of infectious diseases, 209(8), 1231-1240 (2014-01-10)
Staphylococcus aureus is the most common cause of endovascular infections. The staphylococcal accessory regulator A locus (sarA) is a major virulence determinant that may potentially impact methicillin-resistant S. aureus (MRSA) persistence in such infections via its influence on biofilm formation.
G Ralph Corey et al.
The New England journal of medicine, 370(23), 2180-2190 (2014-06-05)
Oritavancin is a lipoglycopeptide with bactericidal activity against gram-positive bacteria. Its concentration-dependent activity and prolonged half-life allow for single-dose treatment. We conducted a randomized, double-blind trial in which adults with acute bacterial skin and skin-structure infections received either a single
Christopher J Rudnisky et al.
Ophthalmology, 121(4), 835-841 (2013-12-12)
To determine the 8-year incidence of endophthalmitis after cataract surgery and to determine which surgical practices were associated with higher rates of endophthalmitis. Case-control study. A total of 75 318 eyes undergoing cataract extractions, performed by 26 different surgeons at 4 public

Articles

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service