Merck
  • Whole-genome sequencing demonstrates that fidaxomicin is superior to vancomycin for preventing reinfection and relapse of infection with Clostridium difficile.
Whole-genome sequencing demonstrates that fidaxomicin is superior to vancomycin for preventing reinfection and relapse of infection with Clostridium difficile.
The Journal of infectious diseases (2013-11-13)
David W Eyre, Farah Babakhani, David Griffiths, Jaime Seddon, Carlos Del Ojo Elias, Sherwood L Gorbach, Tim E A Peto, Derrick W Crook, A Sarah Walker
ABSTRACT
Whole-genome sequencing was used to determine whether the reductions in recurrence of Clostridium difficile infection observed with fidaxomicin in pivotal phase 3 trials occurred by preventing relapse of the same infection, by preventing reinfection with a new strain, or by preventing both outcomes. Paired isolates of C. difficile were available from 93 of 199 participants with recurrences (28 were treated with fidaxomicin, and 65 were treated with vancomycin). Given C. difficile evolutionary rates, paired samples ≤2 single-nucleotide variants (SNVs) apart were considered relapses, paired samples >10 SNVs apart were considered reinfection, and those 3-10 SNVs apart (or without whole-genome sequences) were considered indeterminate in a competing risks survival analysis. Fidaxomicin reduced the risk of both relapse (competing risks hazard ratio [HR], 0.40 [95% confidence interval {CI}, .25-.66]; P = .0003) and reinfection (competing risks HR, 0.33 [95% CI, 0.11-1.01]; P = .05).
Product Description
Vancomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, meets USP testing specifications
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥85% (Vancomycin B)
Millipore
Vancomycin supplement, suitable for microbiology