Merck
  • Inducible loss of one Apc allele in Lrig1-expressing progenitor cells results in multiple distal colonic tumors with features of familial adenomatous polyposis.
Inducible loss of one Apc allele in Lrig1-expressing progenitor cells results in multiple distal colonic tumors with features of familial adenomatous polyposis.
American journal of physiology. Gastrointestinal and liver physiology (2014-05-17)
Anne E Powell, Gregory Vlacich, Zhen-Yang Zhao, Eliot T McKinley, M Kay Washington, H Charles Manning, Robert J Coffey
ABSTRACT
Individuals with familial adenomatous polyposis (FAP) harbor a germline mutation in adenomatous polyposis coli (APC). The major clinical manifestation is development of multiple colonic tumors at a young age due to stochastic loss of the remaining APC allele. Extracolonic features, including periampullary tumors, gastric abnormalities, and congenital hypertrophy of the retinal pigment epithelium, may occur. The objective of this study was to develop a mouse model that simulates these features of FAP. We combined our Lrig1-CreERT2/+ mice with Apcfl/+ mice, eliminated one copy of Apc in leucine-rich repeats and immunoglobulin-like domains protein 1 (Lrig1)-positive (Lrig1(+)) progenitor cells with tamoxifen injection, and monitored tumor formation in the colon by colonoscopy and PET. Initial loss of one Apc allele in Lrig1(+) cells results in a predictable pattern of preneoplastic changes, culminating in multiple distal colonic tumors within 50 days of induction, as well as the extracolonic manifestations of FAP mentioned above. We show that tumor formation can be monitored by noninvasive PET imaging. This inducible stem cell-driven model recapitulates features of FAP and offers a tractable platform on which therapeutic interventions can be monitored over time by colonoscopy and noninvasive imaging.
Product Description
Tropicamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tropicamide, solid
Sigma-Aldrich
Tamoxifen, ≥99%
Supelco
Tamoxifen, analytical standard