Skip to Content
Merck
All Photos(1)

Key Documents

T1022

Supelco

(±)Thiopental solution

1.0 mg/mL in methanol, analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

Assay

≥98%

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

CCCC(C)C1(CC)C(=O)NC(=S)NC1=O

InChI

1S/C11H18N2O2S/c1-4-6-7(3)11(5-2)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)

InChI key

IUJDSEJGGMCXSG-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Lindgren et al.
Neurocritical care, 17(3), 367-373 (2012-08-31)
Non-convulsive seizures (NCSZ) can be more prevalent than previously recognized among comatose neuro-intensive care patients. The aim of this study was to evaluate the frequency of NCSZ and non-convulsive status epilepticus (NCSE) in sedated and ventilated subarachnoid hemorrhage (SAH) patients.
Chadi G Abdallah et al.
The journal of ECT, 28(3), 157-161 (2012-08-01)
Studies now provide strong evidence that the N-methyl-D-aspartate receptor antagonist ketamine possesses rapidly acting antidepressant properties. This study aimed to determine if a low dose of ketamine could be used to expedite and augment the antidepressant effects of electroconvulsive therapy
Tark Purtuloğlu et al.
The journal of ECT, 29(1), 37-40 (2013-02-05)
To compare propofol and sodium thiopental as anesthetic agents for electroconvulsive therapy (ECT) in major depression with respect to clinical effect. Participants were composed of 96 patients with depression who were administered either propofol or sodium thiopental as an anesthetic
Pre-filled thiopental syringes reduce cost and wastage whilst improving safety.
H Murdoch et al.
International journal of obstetric anesthesia, 21(4), 384-385 (2012-08-25)
Ryohei Okazaki et al.
Life sciences, 92(4-5), 259-265 (2013-01-22)
In this study, we investigated the combined effect of Neurotropin® and pregabalin for L5-spinal nerve ligation (L5-SNL) model in rats and thiopental-induced sleep in mice. The left fifth lumbar nerve of rats was tightly ligated with silk sutures under pentobarbital

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service