Skip to Content
Merck
All Photos(1)

Key Documents

S0100000

Salbutamol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

α-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-α,α′-diol, Albuterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21NO3
CAS Number:
Molecular Weight:
239.31
Beilstein:
6405698
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

albuterol, salbutamol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)NCC(O)c1ccc(O)c(CO)c1

InChI

1S/C13H21NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,12,14-17H,7-8H2,1-3H3

InChI key

NDAUXUAQIAJITI-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salbutamol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

β2-adrenoceptor agonist

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Reza Maleki-Yazdi et al.
Respiratory medicine, 108(12), 1752-1760 (2014-12-03)
Combinations of inhaled long-acting bronchodilator therapies such as muscarinic antagonists and β2-agonists may be more effective than monotherapy in the treatment of chronic obstructive pulmonary disease (COPD). This study was a 24-week, Phase III, multicenter, randomized, blinded, double-dummy, parallel-group study
L Gnatiuc et al.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease, 19(1), 21-30 (2014-12-19)
Increasing access to essential respiratory medicines and influenza vaccination has been a priority for over three decades. Their use remains low in low- and middle-income countries (LMICs), where little is known about factors influencing use, or about the use of
Thomas Ritz et al.
Chest, 146(5), 1237-1247 (2014-08-15)
Hyperventilation has been associated with adverse effects on lung function, symptoms, and well-being in asthma. We examined whether raising end-tidal CO2 levels (ie, Pco2) compared with slow breathing is associated with improvements in asthma control, including peak flow variability. One
Karim Makhlouf et al.
CNS drugs, 16(1), 1-8 (2002-01-05)
The beta2-adrenergic receptor agonist salbutamol (albuterol) has been used for many years in the treatment of bronchospasm in patients with asthma. In this patient group, salbutamol is a relatively safe and inexpensive drug, and is easy to administer. Within the
The 1990 Lilly Prize Lecture. A way of looking at agonism and antagonism: lessons from salbutamol, salmeterol and other beta-adrenoceptor agonists.
D Jack
British journal of clinical pharmacology, 31(5), 501-514 (1991-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service