Skip to Content
Merck
All Photos(1)

Key Documents

SML2215

Sigma-Aldrich

Aprepitant

≥98% (HPLC)

Synonym(s):

Aprepitant, 3H-1,2,4-Triazol-3-one, 5-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro, 5-[[(2R,3S)-2-[(1R)-1-[3,5bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3one, Emend, L-754030, MK 869, MK-0869, ONO 7436, 3-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one, 5-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21F7N4O3
CAS Number:
Molecular Weight:
534.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +61 to +71°, c = 1.0 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=CC=C([C@@H]([C@@H](O[C@H](C)C2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2)OCC3)N3CC4=NNC(N4)=O)C=C1

InChI

1S/C23H21F7N4O3/c1-12(14-8-15(22(25,26)27)10-16(9-14)23(28,29)30)37-20-19(13-2-4-17(24)5-3-13)34(6-7-36-20)11-18-31-21(35)33-32-18/h2-5,8-10,12,19-20H,6-7,11H2,1H3,(H2,31,32,33,35)/t12-,19-,20+/m1/s1

InChI key

ATALOFNDEOCMKK-BYYRLHKVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aprepitant has been used as neurokinin 1 (NK1) receptor antagonist in dorsal root ganglion (DRG) derived cells and bone marrow mesenchymal stem cells (BMSCs) coculture.

Biochem/physiol Actions

Aprepitant is an antiemetic drug. It is a moderate cytochrome P450 3A4 (CYP3A4) inhibitor. It is capable of crossing the blood brain barrier in humans. In animals, aprepitant can cross the placental barrier. Aprepitant is a potent and selective antagonist of human substance P/neurokinin 1 (NK1) receptors used clinically to treat chemotherapy-induced or post-operative nausea and vomiting, and being investigated for other activity including possible treatment for depression and pruritis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepti Jacob et al.
American journal of physiology. Gastrointestinal and liver physiology, 313(5), G505-G510 (2017-08-18)
Aprepitant, an NK1 receptor antagonist, is approved for the treatment of chemotherapy-induced or postoperative emesis by blocking NK1 receptors in the brain stem vomiting center. The effects of NK1 receptors on gastric functions and postprandial symptoms in humans are unclear;
Aprepitant and fosaprepitant drug interactions: a systematic review
Patel P, et al.
British Journal of Clinical Pharmacology, 83(10), 2148-2148 (2017)
The role of neurokinin-1 (substance P) antagonists in the prevention of postoperative nausea and vomiting
Okafor D, et al.
Journal of Anaesthesiology, Clinical Pharmacology, 33(4), 441-441 (2017)
Dorsal Root Ganglion Maintains Stemness of Bone Marrow Mesenchymal Stem Cells by Enhancing Autophagy through the AMPK/mTOR Pathway in a Coculture System
Zhang S, et al.
Stem Cells International (2018)
Aprepitant: A substance P antagonist for chemotherapy induced nausea and vomiting
Girish C, et al.
Indian Journal of Cancer, 44(1), 25-25 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service