PHR8818
Zolpidem tartrate
certified reference material, pharmaceutical secondary standard
Synonym(s):
Ambien, Intermezzo, Stilnox
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1724907
API family
zolpidem
packaging
pkg of 500MG
application(s)
pharmaceutical
storage temp.
-10 to -25°C
SMILES string
[n]21c(nc(c2CC(=O)N(C)C)c3ccc(cc3)C)ccc(c1)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O
InChI
1S/C19H21N3O.C4H6O6/c1-13-5-8-15(9-6-13)19-16(11-18(23)21(3)4)22-12-14(2)7-10-17(22)20-19;5-1(3(7)8)2(6)4(9)10/h5-10,12H,11H2,1-4H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1
InChI key
NYVVVBWEVRSKIU-LREBCSMRSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service