Skip to Content
Merck
All Photos(1)

Key Documents

PHR1848

Supelco

Cephalexin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Cephalexin monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O4S · H2O
CAS Number:
Molecular Weight:
365.40
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 61
traceable to Ph. Eur. C0675000
traceable to USP 1099008

API family

cephalexin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

InChI

1S/C16H17N3O4S.H2O/c1-8-7-24-15-11(14(21)19(15)12(8)16(22)23)18-13(20)10(17)9-5-3-2-4-6-9;/h2-6,10-11,15H,7,17H2,1H3,(H,18,20)(H,22,23);1H2/t10-,11-,15-;/m1./s1

InChI key

AVGYWQBCYZHHPN-CYJZLJNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Cephalexin is a cephalosporine which is categorized under the class of β-lactam antibiotics. It is utilized for regulating the effects of Gram-positive and Gram-negative bacteria. It favors the lysis of bacterial cell wall by inhibiting the synthesis of essential structural components. Cephalexin belongs to the safest group of broad-spectrum bactericidal antimicrobial agents.

Application

Cephalexin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and water samples by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0286 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of multiple pharmaceutical classes in surface and ground waters by liquid chromatography-ion trap-tandem mass spectrometry.
Grujic S, et al.
Journal of Chromatography A, 1216(25), 4989-5000 (2009)
Comparison of several methods used for the determination of cephalosporins. Analysis of cephalexin in pharmaceutical samples.
Martinez LG, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(3), 405-423 (2002)
Application of thin layer and high performance liquid chromatography to the separation and determination of cephalexin in cephradine, in bulk powder and in pharmaceuticals.
Crombez E, et al.
Chromatographia, 11(11), 653-657 (1978)
A validated stability indicating HPTLC method for determination of cephalexin in bulk and pharmaceutical formulation.
Jeswani RM, et al.
International Journal of PharmTech Research, 1(3), 527-536 (2009)
Rapid and sensitive high-performance liquid chromatographic determination of four cephalosporin antibiotics in pharmaceuticals and body fluids.
Samanidou VF, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 788(1), 147-158 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service