Skip to Content
Merck
All Photos(1)

Key Documents

32472

Supelco

Cefquinome sulfate

VETRANAL®, analytical standard

Synonym(s):

(6R,7R)-7-[[(2Z)-2-(2-Amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-3-(5,6,7,8-tetrahydroquinolin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate sulfate, Cefquinome sulphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24N6O5S2 · H2SO4
CAS Number:
Molecular Weight:
626.68
Beilstein:
11333438
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS([O-])(=O)=O.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(C[n+]3cccc4CCCCc34)=C(N2C1=O)C(O)=O)c5csc(N)n5

InChI

1S/C23H24N6O5S2.H2O4S/c1-34-27-16(14-11-36-23(24)25-14)19(30)26-17-20(31)29-18(22(32)33)13(10-35-21(17)29)9-28-8-4-6-12-5-2-3-7-15(12)28;1-5(2,3)4/h4,6,8,11,17,21H,2-3,5,7,9-10H2,1H3,(H3-,24,25,26,30,32,33);(H2,1,2,3,4)/b27-16-;/t17-,21-;/m1./s1

InChI key

KYOHRXSGUROPGY-OFNLCGNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cefquinome sulfates are veterinary, parenteral, fourth-generation cephalosporins belonging to the aminothiazolyl class of compounds. They exhibit broad-spectrum antimicrobial activity against gram-positive and gram-negative bacteria such as Pseudomonas aeruginosa and Enterobacteriaceae.

Application

Cefquinome sulfate may be used as an analytical reference standard for the determination of the analyte in:
  • Pharmaceutical dosage form by reversed phase high performance liquid chromatography (RP-HPLC).


It may be used as a precursor for the preparation of cefquinome that can be employed as an analytical reference standard for the analysis of the analyte in:
  • Bovine milk as well as muscle and kidney samples by HPLC and LC coupled to tandem mass spectrometry (MS/MS), respectively.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Smulski et al.
BMC veterinary research, 16(1), 99-99 (2020-04-02)
The purpose of the study described here was to evaluate the effects of different supportive treatments - such as antioxidants, immunomodulators, and nonsteroidal anti-inflammatory drugs (NSAIDs) - in mastitic cows treated with intramammary antibiotics on the efficacy of mastitis therapy
Dongbo Li et al.
Colloids and surfaces. B, Biointerfaces, 182, 110323-110323 (2019-07-20)
Staphylococcus aureus (S. aureus) has the propensity to form biofilms, which eventually cause antibiotic resistance and treatment failure. Cefquinome sulfate (CS) is an animal-specific antibacterial agent for S. aureus infection. In this work, CS cationic proliposomes (CSCPs) were prepared by
Residue analysis of 15 penicillins and cephalosporins in bovine muscle, kidney and milk by liquid chromatography-tandem mass spectrometry.
Becker M, et al.
Analytica Chimica Acta, 520(1-2), 19-32 (2004)
Ivana G Castilho et al.
Journal of dairy science, 100(8), 6414-6421 (2017-06-03)
Staphylococcus aureus is a common pathogen that causes subclinical bovine mastitis due to several virulence factors. In this study, we analyzed S. aureus isolates collected from the milk of cows with subclinical mastitis that had 8 possible combinations of bap
Aleksandar Siljanoski et al.
The Journal of dairy research, 85(3), 321-326 (2018-07-04)
The purpose of this study was to determine tetracycline and other antimicrobial residues in milk from dairy cows treated for clinical mastitis. Data on cow health and drugs used were obtained from the farm veterinarians. A milk sample from each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service