Skip to Content
Merck
All Photos(1)

Key Documents

PHR1483

Supelco

Ferrous Sulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Iron(II) sulfate heptahydrate, Ferrous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1270355

vapor pressure

14.6 mmHg ( 25 °C)

API family

ferrous sulfate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

density

1.898 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ferrous sulfate is the sulfate salt of divalent cation form of iron. It is an essential body mineral, that is commonly used in the treatment of iron deficiency anemia.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Ferrous sulfate may be used as a reference standard for the determination of the analyte in bulk drug and pharmaceutical formulation by titrimetry[2] and spectroscopy.[3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3031 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Delompré et al.
Foods (Basel, Switzerland), 9(10) (2020-10-18)
Orally Disintegrating Tablets (ODTs) are used to restore the nutritional status of people suffering from swallowing pathologies such as dysphagia. ODTs are consumed without water intake and are easily swallowed. The main active compounds of ODTs are vitamins and minerals.
Determination of the iron state in ferrous iron containing vitamins and dietary supplements: application of Mossbauer spectroscopy
Oshtrakh MI, et al.
Journal of Pharmaceutical and Biomedical Analysis, 40(5), 1281-1287 (2006)
Iron-deficiency anemia
Camaschella C
The New England Journal of Medicine, 372(19), 1832-1843 (2015)
Ferrous sulfate
USP43/NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 40(5)(19), 1862-1862 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service