Skip to Content
Merck
All Photos(1)

Key Documents

82950

Supelco

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt

for spectrophotometric det. of Fe, 97.0%

Synonym(s):

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4′,4″-disulfonic acid sodium salt, PDT disulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13N4NaO6S2
CAS Number:
Molecular Weight:
492.46
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

97.0%

form

solid

quality

for spectrophotometric det. of Fe

technique(s)

UV/Vis spectroscopy: suitable

impurities

~7% water

mp

≥300 °C

cation traces

Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Mo: ≤50 mg/kg
Ni: ≤50 mg/kg
V: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].OS(=O)(=O)c1ccc(cc1)-c2nnc(nc2-c3ccc(cc3)S([O-])(=O)=O)-c4ccccn4

InChI

1S/C20H14N4O6S2.Na/c25-31(26,27)15-8-4-13(5-9-15)18-19(14-6-10-16(11-7-14)32(28,29)30)23-24-20(22-18)17-3-1-2-12-21-17;/h1-12H,(H,25,26,27)(H,28,29,30);/q;+1/p-1

InChI key

ZGVNYCXXBQPDPQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt also known as Ferrozine is a chelating agent for ferrous iron.

Application

It was used in determining of chelating activity of Fe2+ using Boyer and McCleary method.

Other Notes

Highly specific water-soluble reagent for the accurate determination of submicrogram levels of serum iron (pH 3.5-4.5, lmax 562 nm, A 29′000)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.L. Stookey
Analytical Chemistry, 42, 779-779 (1970)
Molecular characterization of a putative Arabidopsis thaliana copper transporter and its yeast homologue.
Kampfenkel, Karlheinz, et al.
The Journal of Biological Chemistry, 270.47, 28479-28486 (1995)
Antioxidative activity of protein hydrolysate from round scad muscle using alcalase and flavourzyme.
Thiansilakul, Yaowapa, Soottawat Benjakul, and Fereidoon Shahidi.
Journal of Food Biochemistry, 31.2 , 266-287 (2007)
Direct and indirect assays of serum iron with a sensitive new color reagent.
D Plaut et al.
The American journal of medical technology, 38(3), 82-89 (1972-03-01)
F Ceriotti et al.
Clinical chemistry, 26(2), 327-331 (1980-02-01)
Serum iron is released from transferrin and reduced at pH 1.7 by treating serum with a 10 g/L ascorbic acid solution in 0.1 mol/L HCl. When ferrozine is added to this reagent, it forms a complex with iron that is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service