Skip to Content
Merck
All Photos(1)

Key Documents

PHR1343

Supelco

Sulfadiazine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sulfadiazine, 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide, N1-(Pyrimidin-2-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N4O2S
CAS Number:
Molecular Weight:
250.28
Beilstein:
6733588
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. S1800000
traceable to USP 1625009

API family

sulfadiazine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

253 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ncccn2

InChI

1S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7H,11H2,(H,12,13,14)

InChI key

SEEPANYCNGTZFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfadiazine is a bacteriostatic drug, that belongs to the class of medication referred as sulfonamide antibiotics. It is generally used in the treatment of cerebral meningitis, pneumonia, staphylococcal and streptococcal sepsis.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Sulfadiazine may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by high performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3953 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

33 - Antimicrobial Drugs
Synthesis of Essential Drugs, 499-523 (2006)
Determination of the content and the related substances of silver sulfadiazine gel by HPLC
Li Y, et al.
Chinese Journal of Hospital Pharmacy, 24(8), 474-474 (2004)
Sulfadiazine
Pharmacopeia, US
United States Pharmacopeia/National Formulary, 4106-4106 (2018)
Development and validation of a stability-indicating HPLC method for the simultaneous determination of sulfadiazine sodium and trimethoprim in injectable solution formulation
Ghanem MM and Abu-Lafi SA
Scientia Pharmaceutica, 81(1), 167-182 (2013)
Sulfadiazine
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 3674-3675 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service